Skip to Content
Merck

SML1279

LLY-507

≥97% (HPLC)

Synonym(s):

5-Cyano-2′-{4-[2-(3-methyl-1H-indol-1-yl)ethyl]piperazin-1-yl}-N-[3-(pyrrolidin-1-yl)propyl]biphenyl-3-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H42N6O
CAS Number:
Molecular Weight:
574.76
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC(C1=C2C=CC=C1)=CN2CCN(CC3)CCN3C4=CC=CC=C4C5=CC(C#N)=CC(C(NCCCN6CCCC6)=O)=C5

InChI

1S/C36H42N6O/c1-28-27-42(34-11-4-2-9-32(28)34)22-19-40-17-20-41(21-18-40)35-12-5-3-10-33(35)30-23-29(26-37)24-31(25-30)36(43)38-13-8-16-39-14-6-7-15-39/h2-5,9-12,23-25,27H,6-8,13-22H2,1H3,(H,38,43)

InChI key

PNYRDVBFYVDJJI-UHFFFAOYSA-N

Biochem/physiol Actions

LLY-507 is a potent and selective inhibitor of SMYD2 protein lysine methyltransferase (PKMT) with an in vitro IC50 <15 nM and >100-fold selectivity over other methyltransferases and other non-epigenetic targets. LY-507 has been shown to inhibit p53K370 monomethylation in cells with an IC50 ~600 nM. For full characterization details, please visit the LLY-507 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
LLY-507 is a potent and selective inhibitor of SMYD2 protein lysine methyltransferase (PKMT).

Features and Benefits

LLY-507 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

LLY-507 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the LLY-507 probe summary on the Chemical Probes Portal website.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library