Skip to Content
Merck

A5605

Amlodipine besylate

≥98% (HPLC), L-type calcium channel blocker, powder

Synonym(s):

2-[(2-Aminoethoxy)-methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5- pyridinedicarboxylic acid 3-ethyl 5-methyl ester benzene sulfonate, Norvasc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H25ClN2O5 · C6H5SO3H
CAS Number:
Molecular Weight:
567.05
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Amlodipine besylate, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: ≥20 mg/mL

SMILES string

OS(=O)(=O)c1ccccc1.CCOC(=O)C2=C(COCCN)NC(C)=C(C2c3ccccc3Cl)C(=O)OC

InChI

1S/C20H25ClN2O5.C6H6O3S/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;7-10(8,9)6-4-2-1-3-5-6/h5-8,17,23H,4,9-11,22H2,1-3H3;1-5H,(H,7,8,9)

InChI key

ZPBWCRDSRKPIDG-UHFFFAOYSA-N

Gene Information

Application

Amlodipine besylate is a L-type calcium channel blocker which may be used for the treatment of angina pectoris and hypertension. Amlodipine also inhibits growth of human epidermoid carcinoma A431 cells and has antireproductive effects in male rats.

Biochem/physiol Actions

Amlodipine besylate is an L-type calcium channel blocker.
Amlodipine is an L-type calcium channel blocker. Amlodipine belongs to a class of cardiovascular drugs, which act at the voltage gated calcium channel of the CaV1, or L-type, class. Amlodipine also has antihypertensive and antianginal effects. Its activity resides mainly in the (-)-isomer. Amlodipine inhibits growth of human epidermoid carcinoma A431 cells and has antireproductive effects in male rats.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


Still not finding the right product?

Explore all of our products under Amlodipine besylate


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library