Skip to Content
Merck
All Photos(1)

Key Documents

P0064

Sigma-Aldrich

PalGly

≥98% (HPLC), solid

Synonym(s):

N-(1-Oxohexadecyl)glycine, N-Hexadecanoylglycine, Hexadecanoylaminoacetic acid, N-palmitoyl glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H35NO3
CAS Number:
Molecular Weight:
313.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O=C(CCCCCCCCCCCCCCC)NCC(O)=O

InChI

1S/C18H35NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17(20)19-16-18(21)22/h2-16H2,1H3,(H,19,20)(H,21,22)

InChI key

KVTFEOAKFFQCCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

PalGly is an endogenous signaling lipid that modulates calcium influx and nitric oxide production. PalGly inhibits heat-evoked firing of wide dynamic range (WDR) neurons, activates calcium influx in DRG cells and stimulates NO production. Preliminary studies suggest its signaling mechanism involves GPCR-mediated cation channel activation and a Gαi/o-coupled signaling pathway, which is being further investigated.

Features and Benefits

This compound is featured on the Nitric Oxide Synthases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krishna Pratap Ravindranathan et al.
Journal of the American Chemical Society, 128(17), 5786-5791 (2006-04-28)
UV-vis absorbance measurements and associated studies of cytochrome P450 BM-3 in complex with N-palmitoylglycine (NPG) indicate that a conformational change occurs in the active site of the complex where the terminal atoms of the ligand move from a site distant
Heather B Bradshaw et al.
Prostaglandins & other lipid mediators, 89(3-4), 131-134 (2009-04-22)
A large and growing family of over 70 endogenous lipids of the basic structure N-acyl amide has been identified during the last 10 years. Only a few of these lipids have been characterized for biological activity, however, those that have
D C Haines et al.
Biochemistry, 40(45), 13456-13465 (2001-11-07)
Cytochrome P450s constitute a superfamily of enzymes that catalyze the oxidation of a vast number of structurally and chemically diverse hydrophobic substrates. Herein, we describe the crystal structure of a complex between the bacterial P450BM-3 and the novel substrate N-palmitoylglycine
Conformational dynamics of substrate in the active site of cytochrome P450 BM-3/NPG complex: insights from NMR order parameters.
Krishna Pratap Ravindranathan et al.
Journal of the American Chemical Society, 129(3), 474-475 (2007-01-18)
Kristen A Jeffries et al.
Journal of lipid research, 57(5), 781-790 (2016-03-27)
Long-chain fatty acid amides are signaling lipids found in mammals and other organisms; however, details of the metabolic pathways for the N-acylglycines and primary fatty acid amides (PFAMs) have remained elusive. Heavy-labeled precursor and subtraction lipidomic experiments in mouse neuroblastoma

Articles

We offer many products related to nitric oxide synthases for your research needs.

We offer many products related to nitric oxide synthases for your research needs.

We offer many products related to nitric oxide synthases for your research needs.

We offer many products related to nitric oxide synthases for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service