Skip to Content
Merck
All Photos(3)

Key Documents

B103500

Sigma-Aldrich

Butyric acid

≥99%

Synonym(s):

1-Propanecarboxylic acid, Ethylacetic acid, Propylformic acid, n-Butanoic acid, n-Butyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOH
CAS Number:
Molecular Weight:
88.11
Beilstein:
906770
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

3.04 (vs air)

Quality Level

vapor pressure

0.43 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

824 °F

expl. lim.

10 %

technique(s)

HPLC: suitable

refractive index

n20/D 1.398 (lit.)

pH

2 (25 °C, 10 g/L)
3 (20 °C, 10 g/L)

bp

162 °C (lit.)

mp

−6-−3 °C (lit.)

solubility

water: soluble 10 g/L at 20 °C

density

0.964 g/mL at 25 °C (lit.)

SMILES string

CCCC(O)=O

InChI

1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)

InChI key

FERIUCNNQQJTOY-UHFFFAOYSA-N

Gene Information

human ... HDAC1(3065)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Butyric acid, also known as Ethylacetic acid, is a short-chain volatile fatty acid that is commonly used as a precursor in the synthesis of ethyl butyrate and butyl butyrate. [1]

Application

Butyric acid may be used as a starting material in the preparation of 4-heptanone.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Pasqualucci et al.
Nature, 471(7337), 189-195 (2011-03-11)
B-cell non-Hodgkin's lymphoma comprises biologically and clinically distinct diseases the pathogenesis of which is associated with genetic lesions affecting oncogenes and tumour-suppressor genes. We report here that the two most common types--follicular lymphoma and diffuse large B-cell lymphoma--harbour frequent structural
Butyric acid: applications and recent advances in its bioproduction
Ling J, et al.
Biotechnology Advances, 36, 2101-2117 (2018)
4-Heptanone
Davis R, et al.
Organic Syntheses, 47, 75-75 (1967)
Luis Vázquez et al.
Frontiers in bioengineering and biotechnology, 4, 6-6 (2016-02-24)
Problems derived from obesity and overweight have recently promoted the development of fat substitutes and other low-calorie foods. On the one hand, fats with short- and medium-chain fatty acids are a source of quick energy, easily hydrolyzable and hardly stored
Svenja Plöger et al.
Annals of the New York Academy of Sciences, 1258, 52-59 (2012-06-27)
Butyrate production in the large intestine and ruminant forestomach depends on bacterial butyryl-CoA/acetate-CoA transferase activity and is highest when fermentable fiber and nonstructural carbohydrates are balanced. Gastrointestinal epithelia seem to use butyrate and butyrate-induced endocrine signals to adapt proliferation, apoptosis

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. 안녕하세요. Butyric acid (B103500-5mL) Cell treat 사용 여부에 대한 문의 드립니다.

    1 answer
    1. The suitability for cell culture has not been assessed for product B103500. There certainly is a field of research where this product has been used for cell treatment based applications. Please see the citations listed below for more information.

      Íñiguez-Gutiérrez L, Godínez-Méndez LA, Fafutis-Morris M, Padilla-Arellano JR, Corona-Rivera A, Bueno-Topete MR, Rojas-Rejón ÓA, Delgado-Rizo V. Physiological concentrations of short-chain fatty acids induce the formation of neutrophil extracellular traps in vitro. Int J Immunopathol Pharmacol. 2020 Jan-Dec;34:2058738420958949. doi: 10.1177/2058738420958949. PMID: 33373277; PMCID: PMC7783874.

      Juliana Coronel, Sandra Klausing, Christoph Heinrich, Thomas Noll, Alvio Figueredo-Cardero, Leda R. Castilho, Valeric acid supplementation combined to mild hypothermia increases productivity in CHO cell cultivations, Biochemical Engineering Journal, Volume 114, 2016, Pages 101-109, ISSN 1369-703X, https://doi.org/10.1016/j.bej.2016.06.031.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service