Skip to Content
Merck
All Photos(2)

Key Documents

P0051

Sigma-Aldrich

Anti-PINK1 (N-terminal region) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BRPK, Anti-PARK6, Anti-PTEN induced putative kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cell lysate expressing human PINK1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PINK1(65018)

General description

PTEN induced putative kinase 1 (PINK1) is a serine/threonine mitochondrial kinase. The 581 amino acid protein has an amino terminal mitochondrial target sequence, a putative transmembrane domain, a kinase domain and an autophosphorylation-regulating carboxy terminal domain. The gene encoding it is localized on human chromosome 1p36.12.

Application

Anti-PINK1 (N-terminal region) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

PINK1 (PTEN induced putative kinase 1) has been found to protect neurons from stress-induced mitochondrial dysfunction and apoptosis. Genetic studies in Drosophila indicate that PINK1 acts upstream of Parkin in a common pathway that influences mitochondrial morphology. PINK1 elicits protection in mouse primary neurons from the dopaminergic neurotoxin 1-methyl-4-phenylpyridine (MPP+)/1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) both in vitro and in vivo. In response to enhanced proteasomal stress in vitro, PINK1 has been shown to be cleaved and localized to the mitochondria, and this correlates with increased expression of the processed PINK1 protein in Parkinson′s disease(PD) brain.
PTEN induced putative kinase 1 (PINK1) has a role in removing damaged mitochondria from cells. Mitochondrial damage triggers the accumulation of the protein in the outer mitochondrial membrane, wherein it undergoes autophosphorylation and dimerizes into a supermolecular protein complex. PINK1 then phosphorylates ubiquitin and tags damaged mitochondria for mitophagy. It negatively modulates glioblastoma growth. Mutations in the PINK1 gene have been associated with recessive, early-onset Parkinson′s disease. The protein is expressed in cancerous cells and has a role in cell survival.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of PINK1 Function Promotes Mitophagy through Effects on Oxidative Stress and Mitochondrial Fission*
Ruben K
The Journal of Biological Chemistry (2009)
High expression of PINK1 promotes proliferation and chemoresistance of NSCLC.
Zhang R
Oncology Reports (2017)
Acute focal brain damage alters mitochondrial dynamics and autophagy in axotomized neurons
Cavallucci V, et al.
Cell Death & Disease, 5(11), e1545-e1545 (2014)
PINK1 Is a Negative Regulator of Growth and the Warburg Effect in Glioblastoma.
Agnihotri S
Cancer Research (2016)
Update on Genetics of Parkinsonism
Shinsuke Fujioka Zbigniew K. Wszolek
Neurogenetics (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service