Skip to Content
Merck
All Photos(1)

Key Documents

575001

Sigma-Aldrich

α-Synuclein, Human, Recombinant, E. coli

Synonym(s):

NACP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

suitability

suitable for molecular biology

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SNCA(6622)

General description

Research area: Neuroscience

Recombinant, human α-synuclein expressed in E. coli. α-Synuclein is an acidic neuronal protein of 140 amino acids (amino acid residues 1 -140).

Application

α-Synuclein, Human, Recombinant, E. coli has been used to enhance the filter trap assay for identification of aggregated alpha-synuclein in brain samples.

Biochem/physiol Actions

α-Synuclein been implicated in the pathogenesis of Parkinson’s disease and related neurodegenerative disorders, and more recently, to be an important regulatory component of vesicular transport in neuronal cells.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 20 mM Tris-HCl, 1 mM MgCl₂, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Kahle, P.J, et al. 2001. Am. J. Pathol.159, 2215.
Kim, J. 1997. Mol. Cells7, 78.
Paik, S.R. 1997. Arch. Biochem. Biophys.344, 325.
Jakes, R., et al. 1994. FEBS Lett.345, 27.
Ueda, K., et al. 1993. Proc. Natl. Acad. Sci. USA90, 11282.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimizing filter trap assay for the detection of aggregated alpha-synuclein in brain samples.
Thibauld Oullier et al.
Free neuropathology, 1 (2020-04-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service