Skip to Content
Merck
All Photos(2)

Key Documents

O6014

Sigma-Aldrich

Anti-O-GlcNAc Transferase (TI-14) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-O-linked N-Acetylglucosamine Transferase, Anti-OGT

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 110 kDa

species reactivity

mouse, human, rat

technique(s)

indirect immunofluorescence: 1:50-1:100 using A549 human lung carcinoma cells fixed with paraformaldehyde/triton
western blot: 1:1,000-1:2,000 using HeLa cell nuclear extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OGT(8473)
mouse ... Ogt(108155)
rat ... Ogt(26295)

Related Categories

General description

O-GlcNAc transferase (OGT) catalyzes the addition of an N-acetylglucosamine residue to the amino acids serine or threonine. The enzyme is an homotrimer consisting of three subunits of 110 kDa each, with multiple tetratricopeptide (TPR) repeats.

Immunogen

synthetic peptide corresponding to amino acids 1024-1037 of human O-GlcNAc transferase, conjugated to KLH via an N-terminal added cysteine residue.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

O-linked β-N-acetyl glycosamine (O-GlcNAc) is involved in repressing transcription. O-GlcNAc transferase (OGT) interacts with a histone deacetylase complex by binding to the co repressor Sin3A. This interaction leads to the repression of transcription, after transcription factors and RNA polymerase II are modified by addition of O-GlcNAc. O-GlcNAc modification reversibly inhibits proteosomal function in an ubiquitin-independent fashion.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Berthier et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(47), E11033-E11042 (2018-11-07)
The nuclear receptor REV-ERBα integrates the circadian clock with hepatic glucose and lipid metabolism by nucleating transcriptional comodulators at genomic regulatory regions. An interactomic approach identified O-GlcNAc transferase (OGT) as a REV-ERBα-interacting protein. By shielding cytoplasmic OGT from proteasomal degradation
Sadia Raab et al.
Oncology letters, 23(4), 105-105 (2022-03-05)
Tumor occurrence and development are closely related to metabolism abnormalities. One of the metabolic networks that is dysregulated during carcinogenesis is the fatty acid synthesis pathway, which is mainly controlled by fatty acid synthase (FASN). We previously demonstrated in proliferating
Stéphanie Olivier-Van Stichelen et al.
Frontiers in genetics, 5, 256-256 (2014-08-20)
O-GlcNAc Transferase (OGT) catalyzes protein O-GlcNAcylation, an abundant and dynamic nuclear and cytosolic modification linked to epigenetic regulation of gene expression. The steady-state levels of O-GlcNAc are influenced by extracellular glucose concentrations suggesting that O-GlcNAcylation may serve as a metabolic
O-GlcNAc turns twenty: functional implications for post-translational modification of nuclear and cytosolic proteins with a sugar
Wells L and Hart GW
Febs Letters, 546(1), 154-158 (2003)
Reciprocity between O-GlcNAc and O-phosphate on the carboxyl terminal domain of RNA polymerase II
Comer FI and Hart GW
Biochemistry, 40(26), 7845-7852 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service