Skip to Content
Merck
All Photos(1)

Key Documents

O003

Sigma-Aldrich

β-Funaltrexamine hydrochloride

solid

Synonym(s):

β-FNA hydrochloride, (E)-4-[[(5α,6β)-17-(Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-yl]amino]-4-oxo-2-butenoic acid methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N2O6 · HCl
CAS Number:
Molecular Weight:
490.98
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated

color

white

solubility

H2O: 7.5 mg/mL (aqueous solutions should be promptly used)
methanol: 7.6 mg/mL (do not store in ethanolic solution; methanolic solutions may be stored for several weeks at 4 °C)

storage temp.

−20°C

SMILES string

Cl.COC(=O)\C=C\C(=O)N[C@@H]1CC[C@@]2(O)[C@H]3Cc4ccc(O)c5OC1[C@]2(CCN3CC6CC6)c45

InChI

1S/C25H30N2O6.ClH/c1-32-20(30)7-6-19(29)26-16-8-9-25(31)18-12-15-4-5-17(28)22-21(15)24(25,23(16)33-22)10-11-27(18)13-14-2-3-14;/h4-7,14,16,18,23,28,31H,2-3,8-13H2,1H3,(H,26,29);1H/b7-6+;/t16-,18-,23+,24+,25-;/m1./s1

InChI key

BIPHUOBUKMPSQR-NQGXHZAGSA-N

Biochem/physiol Actions

Selective irreversible μ opioid receptor antagonist that is also a κ opioid receptor agonist.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Cui et al.
Neuroscience, 171(1), 134-143 (2010-09-10)
Hippocampus is a critical structure for the acquisition of morphine-induced conditioned place preference (CPP), which is a usual learning paradigm for assessing drug reward. However, the precise mechanisms remain largely unknown. Phosphatidylinositol 3-kinase (PI3K) and its downstream targets, including Akt
Qing-Zhen Yang et al.
Neuroscience letters, 489(2), 131-135 (2010-12-15)
In the present study, we investigated the antidepressive activity of opiorphin with central administration in the forced swim test in mice. Intracerebroventricular (i.c.v.) administration of opiorphin (1-6 μg/mouse) dose-dependently decreased the immobility time, which was reversed by nonselective opioid receptor
Andrea Bedini et al.
Peptides, 31(11), 2135-2140 (2010-08-18)
We previously described a novel cyclic endomorphin-1 analog c[Tyr-D-Pro-D-Trp-Phe-Gly] (c[YpwFG]), acting as a mu-opioid receptor (MOR) agonist. This study reports that c[YpwFG] is more lipophilic and resistant to enzymatic hydrolysis than endomorphin-1 and produces preemptive antinociception in a mouse visceral
Ai-min Yang et al.
Peptides, 32(9), 1893-1901 (2011-08-16)
Neuronostatin, a recently discovered endogenous bioactive peptide, was encoded by pro-mRNA of somatostatin that contributes to modulation of nociception. However, nociceptive effect of neuronostatin is still not fully known. The aim of this study was to evaluate effect of neuronostatin
A C Shin et al.
Neuroscience, 170(2), 580-588 (2010-07-27)
Overindulgence in easily available energy-dense palatable foods is thought to be an important factor in the current obesity epidemic but the underlying neural mechanisms are not well understood. Here we demonstrate that mu-opioid receptor signaling in the nucleus accumbens may

Protocols

LC/MS Analysis of Opioid Glucuronide Metabolites in Urine on Ascentis® Express F5 after Solid Phase Extraction (SPE) using Supel™-Select HLB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service