Skip to Content
Merck
All Photos(1)

Key Documents

NP10

Sigma-Aldrich

TERGITOL

Type NP-10

Synonym(s):

Mono(p-nonylphenyl)ether, Polyethylene glycol mono(branched p-nonylphenyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

description

non-ionic

Quality Level

type

Type NP-10

mol wt

682 g/mol

CMC

55 ppm

transition temp

cloud point 63 °C

HLB

13.2

SMILES string

CCCCCCCCCC1=CC=C(C=C1)OCCO

InChI

1S/C17H28O2/c1-2-3-4-5-6-7-8-9-16-10-12-17(13-11-16)19-15-14-18/h10-13,18H,2-9,14-15H2,1H3

InChI key

KUXGUCNZFCVULO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tergitol is a nonionic surfactant.

Application

Tergitol has been used in a study to assess the structural characterization and antimicrobial properties of silver nanoparticles. It has also been used in a study to investigate the detection of Salmonella enterica serotype enteritidis in naturally contaminated environmental samples from integrated poultry houses.

Other Notes

Polyglycol ether (nonionic) surfactants.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

476.6 °F - closed cup

Flash Point(C)

247 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohosin Sarkar et al.
Journal of lipids, 2012, 610937-610937 (2012-06-22)
Paraoxonase-1 (PON1) is a serum protein, the activity of which is related to susceptibility to cardiovascular disease and intoxication by organophosphorus (OP) compounds. It may also be involved in innate immunity, and it is a possible lead molecule in the
Carmen Vida et al.
Frontiers in microbiology, 7, 4-4 (2016-02-03)
This study focused on the microbial profile present in an agricultural soil that becomes suppressive after the application of composted almond shells (AS) as organic amendments. For this purpose, we analyzed the functions and composition of the complex communities present
Ning Jie Yang et al.
Reproductive sciences (Thousand Oaks, Calif.), 23(11), 1509-1517 (2016-04-14)
Estrogen-related receptor β (ESRRB), which is a member of the nuclear orphan receptor family, regulates the messenger RNA (mRNA) expression levels of the transcription factors, Oct4 and Nanog, in early embryos and germ cells, thereby maintaining the undifferentiated state and
Rosanna S Kwok et al.
PLoS genetics, 11(7), e1005307-e1005307 (2015-07-02)
Daily rhythms in gene expression play a critical role in the progression of circadian clocks, and are under regulation by transcription factor binding, histone modifications, RNA polymerase II (RNAPII) recruitment and elongation, and post-transcriptional mechanisms. Although previous studies have shown
Ning Jie Yang et al.
Clinical and experimental reproductive medicine, 41(1), 1-8 (2014-04-03)
Estrogen related receptor β (Esrrb) is a member of the orphan nuclear receptors and may regulate the expression of pluripotency-related genes, such as Oct4 and Nanog. Therefore, in the present study, we have developed a method for delivering exogenous ESRRB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service