Skip to Content
Merck
All Photos(1)

Documents

15S9

Sigma-Aldrich

TERGITOL

greener alternative

Type 15-S-9

Synonym(s):

Secondary alcohol ethoxylate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161900
NACRES:
NA.25

description

non-ionic

Quality Level

type

Type 15-S-9

form

liquid

mol wt

596 g/mol

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

52 ppm

transition temp

cloud point 60 °C
pour point 9 °C

HLB

13.3

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Tergitol 15-S-9 is a linear non-ionic surfactant that is a secondary ethoxylated alcohol. This clear liquid surfactant is compatible with anionic, cationic, and other nonionic surfactants.

Application

Tergitol 15-S-9 has been used:
  • To synthesize thermally stable mesoporous alumina
  • To synthesize porous TiO2
  • Along with SDS, ethanol and water for continuous phase preparations in electrospray emulsification
  • In the synthesis of Lath-like γ-alumina and boehmite nanofibers
Tergitol has been used as a surfactant to study its influence on the formation of multi-phase micro-emulsions.

Features and Benefits

  • Superior detergency
  • Rapid dissolution & good rinseability
  • Excellent formulation & handling properties
  • Narrow gel range
  • Readily biodegradable (OECD 301E)
  • Low odor

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

469.4 °F - open cup

Flash Point(C)

243 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rates of solubilization of triolein into nonionic surfactant solutions.
Chen B H, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 128(1-3), 129-143 (1997)
Uniform discotic wax particles via electrospray emulsification.
Journal of Colloid and Interface Science, 334(1), 22-28 (2009)
Preparation method and some tribological properties of porous titanium dioxide layers.
Piwonski I
Thin Solid Films, 515(7-8), 3499-3506 (2007)
Thermally stable mesoporous alumina synthesized with non-ionic surfactants in the presence of amines.
Gonzalez-Pena V, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 44, 203-210 (2001)
Interfacial tension and phase behavior of surfactant-brine?oil system.
Bera A, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 383(1-3), 114-119 (2011)

Related Content

Green chemicals evaluation aligns with the 12 Principles of Green Chemistry, emphasizing resource efficiency and hazard reduction.

Green chemicals evaluation aligns with the 12 Principles of Green Chemistry, emphasizing resource efficiency and hazard reduction.

Green chemicals evaluation aligns with the 12 Principles of Green Chemistry, emphasizing resource efficiency and hazard reduction.

Green chemicals evaluation aligns with the 12 Principles of Green Chemistry, emphasizing resource efficiency and hazard reduction.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service