Skip to Content
Merck
All Photos(1)

Key Documents

N8534

Sigma-Aldrich

Nilutamide

solid

Synonym(s):

5,5-Dimethyl-3-[4-nitro-3-(trifluoromethyl)phenyl]-2,4-imidazolidinedione, Anandron, RU-23908

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F3N3O4
CAS Number:
Molecular Weight:
317.22
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

form

solid

originator

Sanofi Aventis

SMILES string

CC1(C)NC(=O)N(c2ccc(c(c2)C(F)(F)F)[N+]([O-])=O)C1=O

InChI

1S/C12H10F3N3O4/c1-11(2)9(19)17(10(20)16-11)6-3-4-8(18(21)22)7(5-6)12(13,14)15/h3-5H,1-2H3,(H,16,20)

InChI key

XWXYUMMDTVBTOU-UHFFFAOYSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

Application

Nilutamide has been used:
  • as a nuclear androgen receptor (nAR) inhibitor to study its effects on oocyte maturation in zebrafish
  • to determine its effects on bioluminescent Saccharomyces cerevisiae bioreporters in BLYAS assay
  • as a substrate in hydrogenation reaction

Biochem/physiol Actions

Nilutamide is a nuclear androgen receptor inhibitor. It is a nonsteroidal antiandrogen that competitively inhibits the binding of dihydrotestosterone to the androgen receptor. Nilutamide shows a therapeutic effect against prostate cancer.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Desai et al.
Urology, 58(6), 1016-1020 (2001-12-18)
To study the ability of nilutamide to induce prostate-specific antigen (PSA) responses in patients with hormone-resistant prostate cancer who had been exposed to prior antiandrogen therapy, because resistance to antiandrogens may be mediated by altered binding to a mutated or
Ravi A Madan et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 14(14), 4526-4531 (2008-07-17)
We reported previously the first randomized study of any kind in patients with nonmetastatic, castrate-resistant prostate cancer. The study employed vaccine, the hormone nilutamide, and the combined therapy (crossover for each arm) with an endpoint of time to progression. We
Richard Choo et al.
International journal of radiation oncology, biology, physics, 75(4), 983-989 (2009-05-05)
To determine the efficacy of a combined approach of radiotherapy (RT) plus 2-year androgen suppression (AS) as salvage treatment for prostate-specific antigen (PSA) relapse after radical prostatectomy (RP). Seventy-five patients with PSA relapse after RP were treated with salvage RT
Jing Yu et al.
Endocrinology, 151(4), 1822-1828 (2010-03-03)
The mechanisms of testosterone-induced vasodilatation are not fully understood. This study investigated the effect of testosterone on nitric oxide (NO) synthesis and its molecular mechanism using human aortic endothelial cells (HAEC). Testosterone at physiological concentrations (1-100 nm) induced a rapid
Chih-Yang Chang et al.
Fertility and sterility, 85 Suppl 1, 1195-1203 (2006-04-18)
To investigate the effects of androgen and antiandrogen and the expression of androgen receptor on mouse embryonic stem cells (ESCs) and the inner cell mass. Controlled laboratory study. Academic university hospital. Blastocysts from mice developed at the Institute for Cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service