Skip to Content
Merck
All Photos(1)

Key Documents

N5252

Sigma-Aldrich

Nonadecanoic acid

≥98% (GC)

Synonym(s):

C19:0, NSC 11914, Nonadecylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17COOH
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786261
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (GC)

form

powder

mp

68-70 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C19H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21/h2-18H2,1H3,(H,20,21)

InChI key

ISYWECDDZWTKFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Influence of Manure as a Complex Mixture on Soil Sorption of Pharmaceuticals-Studies with Selected Chemical Components of Manure: Nonadecanoic acid is studied for its role in the sorption of pharmaceutical compounds in soil, suggesting its importance in agricultural runoff management and environmental safety (Thiele-Bruhn and Zhang, 2023).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Yu et al.
Animal bioscience, 34(9), 1569-1578 (2021-03-08)
This study was conducted to investigate the potential effects of prolonged photoperiod on the serum lipids, carcass traits, and meat quality of Jinjiang cattle during winter. Thirty-four Jinjiang bulls aged between 14 and 16 months were randomly assigned to two
V Fragua et al.
Journal of animal physiology and animal nutrition, 99 Suppl S1, 48-59 (2015-04-14)
Esterified fatty acid oils (EAOs) are obtained from esterification of vegetable acid oils with glycerol. These fat sources have the same fatty acid (FA) composition as their respective native oils but new chemical properties. Several studies have confirmed the potential
C N Marcos et al.
Journal of dairy science, 103(2), 1472-1483 (2019-11-25)
The use of agroindustrial by-products, such as dried distillers grains with solubles (DDGS) and dried citrus pulp (DCP), has been widely investigated in dairy cows, but information on their effects in dairy goats is limited. The influence of feeding olive
Bhushan V Kulkarni et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 306(12), R879-R885 (2014-04-04)
Lingual lipase generates nonesterified fatty acids (NEFA) from dietary fats during oral processing by lipolysis. Lingual lipase in rodents has strong lipolytic activity and plays a critical role in oral detection of fats. The functional activity of lingual lipase during
Manish Kumar Patel et al.
Bio-protocol, 8(18), e3014-e3014 (2018-09-20)
Fatty acids (FAs) are carboxylic acids with long aliphatic chains that may be straight, branched and saturated or unsaturated. Most of the naturally occurring plant FAs contains an even number of carbon (C4-C24). FAs are used in food and pharmacological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service