Skip to Content
Merck
All Photos(1)

Key Documents

N2126

Sigma-Aldrich

Neostigmine methyl sulfate

Synonym(s):

3-(N,N-Dimethylcarbamoyloxy)-N,N,N,-trimethylanilinium methyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22N2O6S
CAS Number:
Molecular Weight:
334.39
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

storage temp.

−20°C

SMILES string

COS([O-])(=O)=O.CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C

InChI

1S/C12H19N2O2.CH4O4S/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;1-5-6(2,3)4/h6-9H,1-5H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

OSZNNLWOYWAHSS-UHFFFAOYSA-M

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Reversible inhibitor of acetylcholinesterase that is similar to eserine, but does not cross the blood-brain barrier.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C T Sadashiva et al.
Bioorganic & medicinal chemistry letters, 16(15), 3932-3936 (2006-06-01)
The cholinergic hypothesis of Alzheimer's disease (AD) has spurred the development of numerous structural classes of compounds with different pharmacological profile aimed at increasing central cholinergic neurotransmission. Thus proving a symptomatic treatment for this disease are cholinomimetics with the pharmacological
Michael Benatar et al.
The Cochrane database of systematic reviews, 12, CD005081-CD005081 (2012-12-14)
Approximately 50% of people with myasthenia gravis present with purely ocular symptoms, so called ocular myasthenia. Of these between 50% to 60% develop generalized disease, most within two years. Their management is controversial. This is an update of a review
Qian-Sheng Yu et al.
Bioorganic & medicinal chemistry, 18(13), 4687-4693 (2010-07-16)
The N-monophenylcarbamate analogues of neostigmine methyl sulfate (6) and pyridostigmine bromide (8) together with their precursors (5), (7), and the N(1)-methylammonium analogues of (-)-phenserine (12), (-)-tolserine (14), (-)-cymserine (16) and (-)-phenethylcymserine (18) were synthesized to produce long-acting peripheral inhibitors of
Martina Grosse-Sundrup et al.
BMJ (Clinical research ed.), 345, e6329-e6329 (2012-10-19)
To determine whether use of intermediate acting neuromuscular blocking agents during general anesthesia increases the incidence of postoperative respiratory complications. Prospective, propensity score matched cohort study. General teaching hospital in Boston, Massachusetts, United States, 2006-10. 18,579 surgical patients who received
Oyun Khookhor et al.
Neuro endocrinology letters, 34(1), 58-61 (2013-03-26)
We previously demonstrated that the direct microinjection of cholinesterase inhibitor (neostigmine) into the hippocampus in rats activated the hypothalamo-pituitary -adrenal axis and increased the level of norepinephrine in the plasma. In the current study we tried to measure the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service