Skip to Content
Merck
All Photos(1)

Key Documents

MTOXCE19A1

Sigma-Aldrich

CypExpress Aromatase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

biological source

human

recombinant

expressed in Pichia pastoris

form

dry powder

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP19A1(1588)

Biochem/physiol Actions

Aromatase is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and catalyzes the last steps of estrogen biosynthesis. Mutations in this gene can result in either increased or decreased aromatase activity; the associated phenotypes suggest that estrogen functions both as a sex steroid hormone and in growth or differentiation. Alternative promoter use and alternative splicing results in multiple transcript variants that have different tissue specificities
Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous.

Features and Benefits

CypExpress 19A1 also know as Aromatase is a permeabilized and stabilized dried yeast powder preparation containing full length, unmodified, human CYP 19A1 and recombinant human NADPH oxidoreductase. Using proprietary processes, CypExpress are expressed in Pichia yeast. Following optimal expression, the cells are inactivated, permeabilized and dried to a fine powder. CypExpress retains the cellular mechanisms to provide the P450 enzymes with the energy and cofactors to continue to function for long experiments, and can generate larger amounts of metabolite than mammalian microsomes or other genetically engineered expression systems.It is designed for rapid screening of drug metabolites and for scaling up metabolite generation.

Legal Information

CypExpress is a trademark of Oxford Biomedical Research

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asli Sezgin Caglar et al.
Endocrine, 49(3), 761-768 (2015-02-24)
The aim of this study is to evaluate aromatase expression in prolactin (PRL), thyroid stimulating hormone (TSH), and growth hormone (GH) secreting cells. Nontumoral human pituitary specimens were obtained from autopsy samples. Aromatase co-expression was determined by double immunohistochemical staining
Aromatase deficiency: rare cause of virilization.
Barbara Ludwikowski et al.
European journal of pediatric surgery : official journal of Austrian Association of Pediatric Surgery ... [et al] = Zeitschrift fur Kinderchirurgie, 23(5), 418-422 (2012-10-25)
Hong Zhao et al.
Journal of molecular endocrinology, 57(1), R19-R33 (2016-04-14)
Long-term exposure to excess estrogen increases the risk of breast cancer and type 1 endometrial cancer. Most of the estrogen in premenopausal women is synthesized by the ovaries, while extraovarian subcutaneous adipose tissue is the predominant tissue source of estrogen
S C Janicki et al.
Dementia and geriatric cognitive disorders, 35(5-6), 340-346 (2013-05-03)
Few studies of gene variants that affect estrogen activity investigate their association with risk for Alzheimer's disease (AD) in women of different ethnicities. We investigated the influence of CYP19 polymorphisms on risk for AD in a multiethnic cohort of women
Yong Joo Park et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 87, 157-165 (2015-12-27)
Ginkgo biloba has been used in herbal medicines for thousands of years. Although a standard G. biloba extract, EGb 761 has been used to improve cognition in breast cancer patients, its effects on breast cancer are unknown. Therefore, we investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service