Skip to Content
Merck
All Photos(1)

Key Documents

MAK313

Sigma-Aldrich

Adipolysis Assay Kit

sufficient for 200 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

application(s)

pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

endocrinological disorders, diabetes; obesity; cardiovascular diseases; cancer

storage temp.

−20°C

General description

Obesity is a chronic condition that develops from storage of excessive energy in the form of adipose tissue. The resulting adiposity presents a high risk factor for diseases such as type 2 diabetes, cardiovascular diseases, and cancer. Adipolysis or lipolysis is a highly regulated process in fat metabolism, in which triglycerides are broken down into glycerol and free fatty acids. Rapid, robust and accurate procedures for adipolysis quantification in cell culture are very useful in research and drug discovery.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for adipolysis quantification in cell culture.

Principle

This adipolysis assay kit directly measures glycerol released during adipolysis in cultured cells. This homogeneous assay uses a single Working Reagent that combines glycerol kinase, glycerol phosphate oxidase, and color reactions in one step, resulting in a colorimetric (570 nm)/fluorimetric (λex = 530/ λem = 587 nm) product, proportional to the glycerol present.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liliya V Vasileva et al.
International journal of molecular sciences, 21(24) (2020-12-30)
Caffeic acid (CA) and chlorogenic acid (CGA) are phenolic compounds claimed to be responsible for the metabolic effects of coffee and tea consumption. Along with their structural similarities, they share common mechanisms such as activation of the AMP-activated protein kinase
Liliya V Vasileva et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 149, 112002-112002 (2021-01-22)
Chronic low-grade inflammation is a hallmark of obesity and its related metabolic disorders. At the same time signaling from pro-inflammatory factors such as transforming growth factor beta (TGF-β) or interleukin 17A (IL-17A) are proposed as crucial for the commitment of
Saveta G Mladenova et al.
Frontiers in pharmacology, 12, 707507-707507 (2021-09-07)
Obesity is a persistent and continuously expanding social health concern. Excessive fat mass accumulation is associated with increased risk of chronic diseases including diabetes, atherosclerosis, non-alcoholic steatohepatitis, reproductive dysfunctions and certain types of cancer. Alchemilla monticola Opiz. is a perennial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service