MAK119
Acetylcholinesterase Activity Assay Kit
sufficient for 100 colorimetric tests
Synonym(s):
ACHE Activity Assay, RBC Cholinesterase Activity Assay
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
usage
sufficient for 100 colorimetric tests
detection method
colorimetric
relevant disease(s)
neurological disorders
storage temp.
room temp
Gene Information
human ... ACHE(43)
mouse ... ACHE(11423)
Related Categories
General description
Acetylcholinesterases (AChEs) are enzymes that hydrolyze the neurotransmitter acetylcholine (ACh) to acetate and choline. AChE is located at the synaptic cleft and functions to terminate synaptic transmission by catalyzing the breakdown of ACh allowing cholinergic neurons to return to a resting state after activation. It is also found in membranes of red blood cells, motor and sensory fibers, muscles, nerves and central and peripheral tissues. Changes in AChE activity may result from exposure to certain insecticides, which act as cholinesterase inhibitors. Inhibitors of AChE are also used to treat certain conditions such as dementia.
Application
Acetylcholinesterase activity assay kit has been used to determine the activity of acetylcholinesterase in a rat organophosphate model and in brain tissue homogenates.
Features and Benefits
Compatible with high-throughput handling systems. Can be adapted for use with cuvettes.
Suitability
Suitable for the detection of acetylcholinesterase activity in biological samples such as blood, serum, and plasma.
Principle
This assay is an optimized version of the Ellman method in which thiocholine, produced by AChE, reacts with 5,5′-dithiobis(2-nitrobenzoic acid) to form an colorimetric (412 nm) product, proportional to the AChE activity present. One unit of AChE is the amount of enzyme that catalyzes the production of 1.0 μmole of thiocholine per minute at room temperature at pH 7.5. This kit has a linear range of 10-600 units/L of AChE activity.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hemanga Hazarika et al.
Scientific reports, 12(1), 2180-2180 (2022-02-11)
Mosquito-borne infections like dengue, malaria, chikungunya, etc. are a nuisance and can cause profound discomfort to people. Due to the objectional side effects and toxicity associated with synthetic pyrethroids, N,N-diethyl-3-methylbenzamide (DEET), N,N-diethyl phenylacetamide (DEPA), and N,N-di ethyl benzamide (DEBA) based
Abdullah M Alzahrani
Environmental science and pollution research international, 26(16), 16548-16555 (2019-04-14)
Despite the widespread use of the insecticide imidacloprid (IMI), a neonicotinoid, there is an urgent need for documenting information related to its acute toxicity. Therefore, this study aims to explore the markers of IMI acute toxicity in the testes of
Study on social isolation as a risk factor in development of Alzheimer?s disease in rats.
Ali A A, et al.
Brain disorders & therapy., 6(230), 2-2 (2017)
Li-Ping Liang et al.
Toxicological sciences : an official journal of the Society of Toxicology, 162(2), 611-621 (2017-12-23)
Prolonged seizure activity or status epilepticus (SE) is one of the most critical manifestations of organophosphate exposure. Previous studies in our laboratory have demonstrated that oxidative stress is a critical mediator of SE-induced neuronal injury. The goal of this study
Zhen-Xing Zuo et al.
Neural plasticity, 2015, 453170-453170 (2015-12-24)
Chronic pain is a major health issue and most patients suffer from spontaneous pain. Previous studies suggest that Huperzine A (Hup A), an alkaloid isolated from the Chinese herb Huperzia serrata, is a potent analgesic with few side effects. However
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service