Skip to Content
Merck
All Photos(2)

Key Documents

M8515

Sigma-Aldrich

Monastrol

≥98% (HPLC), solid

Synonym(s):

4-(3-Hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-4H-pyrimidin-5-carboxylic Acid Ethyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O3S
CAS Number:
Molecular Weight:
292.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

white to off-white

mp

185-185.9 °C (lit.)

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

CCOC(=O)C1=C(C)NC(=S)NC1c2cccc(O)c2

InChI

1S/C14H16N2O3S/c1-3-19-13(18)11-8(2)15-14(20)16-12(11)9-5-4-6-10(17)7-9/h4-7,12,17H,3H2,1-2H3,(H2,15,16,20)

InChI key

LOBCDGHHHHGHFA-UHFFFAOYSA-N

Application

Monastrol has been used:
  • to treat MDA-MB-231 cells as a non-microtubule-targeting agent
  • as a antineoplastic agent, to treat mouse myeloma cell line SP 2/0, to induce apoptosis and to elucidate the role of metabotropic glutamate receptor 3 (Grm3) in apoptosis
  • as an inhibitor of pteridine reductase in GFP-transfected promastigotes infected macrophages for flow cytometer-based growth inhibition assay and to evaluate anti-leishmanial activity of Leishmania donovani hamster model in vivo

Biochem/physiol Actions

Monastrol is a potent, cell-permeant inhibitor of mitosis. Monastrol arrested cells are characterized by monopolar spindles. This phenotype is induced through specific disruption of mitotic molecular motor kinesin Eg5 with IC50 at 14 μM. No effect on other motor proteins and tubulin.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Packaging

Packaged under inert gas.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabotropic glutamate receptor 3 is involved in B-cell-related tumor apoptosis
Liu X, et al.
International journal of oncology, 49(4), 1469-1478 (2016)
M410, a combretastatin A4 analogue, disrupts microtubules and inhibits HIF-1 alpha in human breast cancer cells
Yang H, et al.
Oncology Reports, 34(1), 334-340 (2015)
T U Mayer et al.
Science (New York, N.Y.), 286(5441), 971-974 (1999-11-05)
Small molecules that perturb specific protein functions are valuable tools for dissecting complex processes in mammalian cells. A combination of two phenotype-based screens, one based on a specific posttranslational modification, the other visualizing microtubules and chromatin, was used to identify
Mayumi Kitagawa et al.
PloS one, 8(6), e64826-e64826 (2013-06-12)
Although Aurora B is important in cleavage furrow ingression and completion during cytokinesis, the mechanism by which kinase activity is targeted to the cleavage furrow and the molecule(s) responsible for this process have remained elusive. Here, we demonstrate that an
Renske van Leuken et al.
PloS one, 4(4), e5282-e5282 (2009-04-25)
Polo-like kinase-1 (Plk1) is activated before mitosis by Aurora A and its cofactor Bora. In mitosis, Bora is degraded in a manner dependent on Plk1 kinase activity and the E3 ubiquitin ligase SCF-betaTrCP. Here, we show that Plk1 is also

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service