Skip to Content
Merck

S5265

Streptolysin O from Streptococcus pyogenes

lyophilized powder, Protein ~3 % by Lowry, 25,000-50,000 U/vial

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
308-500-3
NACRES:
NA.85
MDL number:
Form:
lyophilized powder
Storage condition:
(Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Segment

specific activity

25,000-50,000 U/vial

mol wt

native 69 kDa

composition

Protein, ~3% Lowry

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.)

color

white

solubility

H2O: soluble 1 mg/mL

antibiotic activity spectrum

neoplastics

mode of action

cell membrane | interferes

storage temp.

2-8°C

Gene Information

Streptococcus pyogenes MGAS8232 ... slo(993428)

Application

Permeabilizes membranes to permit cellular uptake of large or charged molecules.
Streptolysin O is a toxin secreted by Streptococcus pyogenes and is a prototype member of poreforming bacterial cytolysins. It is used permeabilize cell membranes to permit cellular uptake of large or charged molecules. It is used to study macromolecule delivery. It is a potential anticancer agent and is used to study suicide cancer gene therapy.
Used to create holes (reversible pores) in cholesterol-enriched plasma membranes of eukaryotic cells allowing selective permeabilization with molecules such as proteins (up to 100 kDa) and siRNAs.

Biochem/physiol Actions

Streptolysin O possesses a single polypeptide chain with a molecular weight of f62 kDa. Streptolysin O binds to membrane cholesterol and oligomerizes to create a ring structure that consists of 45 to 50 units. The ring structure inserts into the membrane to make a large pore (25 to 30 nm), which DNA, RNA, peptides and proteins may pass. It is thiol-activated. It is inhibited by allicin, an active component of garlic.
Thiol-activated toxin that permeabilizes animal cell membranes. The protein binds as a monomer to membrane cholesterol and subsequently polymerizes into large arc- and ring-shaped structures surrounding pores of >12 nm.
Thiol-activated toxin that permeabilizes animal cell membranes..

Packaging

25KU

Physical form

Lyophilized powder containing Tris buffer salts, sodium azide, PMSF, and EDTA.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place
One unit will cause 50% lysis of 2% red blood cell suspensions in phosphate buffered saline, pH 7.4, after incubation at 37 °C for 30 min.


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library