Skip to Content
Merck
All Photos(1)

Key Documents

M7566

Sigma-Aldrich

Monoamine Oxidase Insect Cell Control

recombinant, expressed in wild-type baculovirus infected BTI insect cells

Synonym(s):

MAO negative control

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in wild-type baculovirus infected BTI insect cells

Quality Level

form

liquid

packaging

vial of ~2.5 mg

shipped in

dry ice

storage temp.

−70°C

General description

Monoamine oxidase is a mitochondrial outermembrane flavoenzyme that is a target for antidepressant and neuroprotective drugs.

Application

Monoamine Oxidase inhibitors (MAOIs) are used to treat mental disorders including depression and Parkinson′s.
This product is suitable as a negative control for MAO-A and MAO-B.

Biochem/physiol Actions

Monoamine Oxidase (MAO) is an integral flavoprotein of the outer mitochondrial membrane. MAOs are responsible for catalyzing the oxidative deamination of a wide variety of xenobiotic and endobiotic primary, secondary, and tertiary amines. The primary endogenous function of MAOs involves the inactivation of monoamine neurotransmitters, such as serotonin and dopamine. MAOs exist in two isoforms, MAO-A and MAO-B, which share approximately 70% sequence identity on the amino acid level.1 Both isoforms are nearly ubiquitous in mammals, but show particularly high enzymatic activity in the central nervous system and liver.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molly Wimbiscus et al.
Cleveland Clinic journal of medicine, 77(12), 859-882 (2010-12-15)
Monoamine oxidase (MAO) inhibitors were the first antidepressants introduced, but their use has dwindled because of their reported side effects, their food and drug interactions, and the introduction of other classes of agents. However, interest in MAO inhibitors is reviving.
Interactions of nitrogen-containing xenobiotics with monoamine oxidase (MAO) isozymes A and B: SAR studies on MAO substrates and inhibitors.
A S Kalgutkar et al.
Chemical research in toxicology, 14(9), 1139-1162 (2001-09-18)
M S Benedetti
Fundamental & clinical pharmacology, 15(2), 75-84 (2001-07-27)
Although the cytochrome P450 (CYP) system ranks first in terms of catalytic versatility and the wide range of xenobiotics it detoxifies or activates to reactive intermediates, the contribution of amine oxidases and in particular of monoamine oxidases (MAOs) to the
M B Youdim et al.
Biochemical pharmacology, 41(2), 155-162 (1991-01-15)
Identification, cellular localization, and cDNA cloning of MAO subtypes A and B have increased the insight into the pharmacology of these enzymes, whose primary functions are intra- and extraneuronal inactivation of neurotransmitter (dopamine, noradrenaline and serotonin) and other biogenic amines.
Justin P Wright et al.
European journal of nuclear medicine and molecular imaging, 49(11), 3797-3808 (2022-05-22)
[18F]-labeled positron emission tomography (PET) radioligands permit in vivo assessment of Alzheimer's disease biomarkers, including aggregated neurofibrillary tau (NFT) with [18F]flortaucipir. Due to structural similarities of flortaucipir with some monoamine oxidase A (MAO-A) inhibitors, this study aimed to evaluate flortaucipir

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service