Skip to Content
Merck

Skip To

M2645

Minimum Essential Medium Eagle

HEPES modification, with Earle′s salts and ʟ-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Minimum Essential Medium Eagle, HEPES Modification, with Earle′s salts, L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture

form

powder

technique(s)

cell culture | mammalian: suitable

components

HEPES: 25 mM
phenol red: yes
L-glutamine: yes
NaHCO3: no
sodium pyruvate: no
Earle’s salts (5% CO2): yes

shipped in

ambient

storage temp.

2-8°C

Preparation Note

Formulated to contain 14.2 grams of powder per liter of medium.
Supplement with 2.2 g/L sodium bicarbonate.

Other Notes

Leaving the bicarbonate out gives the investigator greater control over the buffer system.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M0518M8028M1018
description

HEPES Modification, with Earle′s salts, L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture

description

Joklik Modification, with L-glutamine, without calcium chloride and sodium bicarbonate, suitable for cell culture

description

Joklik Modification, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

description

With Hanks′ salts, L-glutamine and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture

form

powder

form

powder

form

liquid

form

powder

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

components

HEPES: 25 mM
phenol red: yes
L-glutamine: yes
NaHCO3: no
sodium pyruvate: no
Earle’s salts (5% CO2): yes

components

HEPES: no
sodium pyruvate: no
L-glutamine: yes
NaHCO3: no
phenol red: yes

components

glucose: yes
phenol red: yes
HEPES: no
Hanks’ salts (2% CO2): no
L-glutamine: no
NaHCO3: yes
Earle’s salts (5% CO2): no
sodium pyruvate: no
stable glutamine: no

components

phenol red: yes
NaHCO3: no
HEPES: no
sodium pyruvate: no
Hanks’ salts (2% CO2): yes
L-glutamine: yes

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

classic media, classical media, FBS

classic media, classical media, FBS, suspension cultures

classic media, classical media, FBS

MEM, classic media, classical media, FBS


Still not finding the right product?

Explore all of our products under Minimum Essential Medium Eagle


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters