Skip to Content
Merck
All Photos(1)

Documents

M8028

Sigma-Aldrich

Minimum Essential Medium Eagle

Joklik Modification, with sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

JMEM, Joklik Modified MEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Minimum Essential Medium Eagle, Joklik Modification, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: yes
phenol red: yes
HEPES: no
Hanks’ salts (2% CO2): no
L-glutamine: no
NaHCO3: yes
Earle’s salts (5% CO2): no
sodium pyruvate: no
stable glutamine: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Recommended for suspension cultures.

Reconstitution

Supplement with 0.292 g/L L-glutamine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangchong Wang et al.
Journal of molecular and cellular cardiology, 104, 63-74 (2017-01-26)
The intracellular fibroblast growth factors (iFGF/FHFs) bind directly to cardiac voltage gated Na
Marcus Rohnke et al.
Journal of controlled release : official journal of the Controlled Release Society, 262, 159-169 (2017-08-02)
Drug functionalization of biomaterials is a modern and popular approach in biomaterials research. Amongst others this concept is used for the functionalization of bone implants to locally stimulate the bone healing process. For example strontium ions (Sr
Gerard A Tarulli et al.
Breast cancer research : BCR, 15(1), R10-R10 (2013-02-02)
The molecular circuitry of different cell types dictates their normal function as well as their response to oncogene activation. For instance, mice lacking the Wip1 phosphatase (also known as PPM1D; protein phosphatase magnesium-dependent 1D) have a delay in HER2/neu (human
Ruiyuan Zhao et al.
Journal of histotechnology, 43(3), 125-134 (2020-04-22)
Adipose-derived mesenchymal stem cells (ADSC) are adult pluripotent cells and important resources for cell-based therapies of animals. There are presently different kinds of somatic cells used as donor cells for clone successfully. However, studies on somatic cell nuclear transplantation (SCNT)
Lei Sheng et al.
The Journal of steroid biochemistry and molecular biology, 189, 240-247 (2019-01-18)
Active vitamin D (1,25(OH)2D) has been shown to regulate numerous cell processes in mammary cells. Degradation of 1,25(OH)2D is initiated by the mitochondrial enzyme, 25-hydroxyvitamin D 24-hydroxylase (CYP24 A1), and provides local control of 1,25(OH)2D bioactivity. Several reports of the association

Articles

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service