Skip to Content
Merck
All Photos(3)

Key Documents

M1255

Sigma-Aldrich

Calcein Blue

Powder

Synonym(s):

4-Methylumbelliferone-8-methyliminodiacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO7
CAS Number:
Molecular Weight:
321.28
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Calcein Blue, fluorescent dye

form

powder

color

white to light yellow

solubility

1 M NaOH: 50 mg/mL

fluorescence

λex 322 nm; λem 445 nm in methanol

suitability

suitable for indicator for EDTA titration

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC1=CC(=O)Oc2c(CN(CC(O)=O)CC(O)=O)c(O)ccc12

InChI

1S/C15H15NO7/c1-8-4-14(22)23-15-9(8)2-3-11(17)10(15)5-16(6-12(18)19)7-13(20)21/h2-4,17H,5-7H2,1H3,(H,18,19)(H,20,21)

InChI key

RJPSHDMGSVVHFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Calcein Blue is a cell-permeable dye that stains viable cells. Upon entering the cell, intracellular esterases cleave the acetoxymethyl (AM) ester group, yielding the membrane-impermeable Calcein Blue fluorescent dye. The staining applications of Calcein blue include tumor tissues, mineralized tissues, bone, amino acids, and many more.

Application

Calcein blue has been used fluorescent labeling of the mineralised sections of bone. Calcein blue AM ester hydrolyzed to calcein blue is suitable as a a viability stain, both microscopically, and in fluorescence activated cell sorting systems. It is also used as a merker of living cells like Schwann cells at neuromuscular junctions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaomian Wu et al.
International journal of nanomedicine, 7, 1215-1225 (2012-03-16)
Compared with titanium (Ti) and other metal implant materials, poly(ether-ether ketone) (PEEK) shows outstanding biomechanical properties. A number of studies have also reported attractive bioactivity for nano-TiO(2) (n-TiO(2)). In this study, n-TiO(2)/PEEK nanocomposites were prepared, taking advantage of the unique
Liangbo Hu et al.
Virologica Sinica, 34(4), 454-466 (2019-06-16)
Baculovirus can transduce a wide range of mammalian cells and is considered a promising gene therapy vector. However, the low transduction efficiency of baculovirus into many mammalian cells limits its practical application. Co-expressing heterologous viral glycoproteins (GPs), such as vesicular
Indicators: International Series of Monographs in Analytical Chemistry (2013)
Zachary J Reitman et al.
Nature communications, 10(1), 3731-3731 (2019-08-21)
Pilocytic astrocytoma (PA), the most common childhood brain tumor, is a low-grade glioma with a single driver BRAF rearrangement. Here, we perform scRNAseq in six PAs using methods that enabled detection of the rearrangement. When compared to higher-grade gliomas, a
Tobias A Fuchs et al.
The Journal of cell biology, 176(2), 231-241 (2007-01-11)
Neutrophil extracellular traps (NETs) are extracellular structures composed of chromatin and granule proteins that bind and kill microorganisms. We show that upon stimulation, the nuclei of neutrophils lose their shape, and the eu- and heterochromatin homogenize. Later, the nuclear envelope

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service