Skip to Content
Merck
All Photos(1)

Key Documents

M0321

Sigma-Aldrich

MDM2, GST-tagged human

recombinant, expressed in E. coli, buffered aqueous glycerol solution, ≥75% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in E. coli

description

contains an N-terminal GST tag

Assay

≥75% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~84 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MDM2(4193)

General description

Murine double minute 2 (MDM2) encoding gene is localized at 12q15 in the human chromosome. It is a multidomain nuclear phosphoprotein. It possesses really interesting new gene (RING) finger domain and its function and structural integrity is required for the action of MDM2. It also has central acidic domain.

Biochem/physiol Actions

MDM2 is a negative regulator of the p53 tumor suppressor.
Mdm2 functions as an E3 ubiquitin ligase and an inhibitor of p53 transcriptional activation.
Murine double minute 2 (MDM2) is a chief regulator of the p53 tumor suppressor protein. It is a multifunctional oncoprotein that contributes to both p53-dependent and -independent roles in oncogenesis. MDM2 has various p53-independent functions that contribute to its activity as an oncogene.
Overexpression of Murine double minute 2 (MDM2) is observed in various cancer like lung, gastric, colorectal, hepatocellular, breast, endometrial, and prostate. A common single nucleotide polymorphism is observed at 309 amino acid (from T to G) increases the susceptibility to cancer.

Physical form

Solution in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF and 25% glycerol.

Preparation Note

Supplied as a truncated version (202-end) with an N-terminal GST tag

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MDM2 promotes proteasome-dependent ubiquitin-independent degradation of retinoblastoma protein
Sdek P, et al.
Molecular Cell, 20(5), 699-708 (2005)
The Mdm2 RING domain C-terminus is required for supramolecular assembly and ubiquitin ligase activity
Poyurovsky MV, et al.
The Embo Journal, 26(1), 90-101 (2007)
[Extracorporeal shock wave lithotripsy (ESWL) in the treatment of urolithiasis].
T Schärfe et al.
Archivos espanoles de urologia, 41(6), 455-459 (1988-07-01)
Fluorescence in situ hybridization for MDM2 gene amplification as a diagnostic tool in lipomatous neoplasms
Weaver J, et al.
Modern Pathology, 21(8), 943-943 (2008)
Association between MDM2 SNP309 and endometrial cancer risk: A PRISMA-compliant meta-analysis
Zou X, et al.
Medicine, 97(49) (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service