Skip to Content
Merck
All Photos(1)

Key Documents

P6249

Sigma-Aldrich

p53 human

recombinant, expressed in E. coli, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

UniProt accession no.

application(s)

genomic analysis

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TP53(7157)

Looking for similar products? Visit Product Comparison Guide

General description

p53 is regulated by MDM2, which acts as a negative regulator of p53 transcription by promoting its degradation by functioning as an E3 ubiquitin ligase which recognizes the N′ terminal transactivation domain.

Application

Useful for the study of postranslational modification of p53, or protein-protein interaction studies.

Biochem/physiol Actions

p53 gene is highly conserved and expressed in normal tissues. It is the most commonly mutated gene in human cancer and more then 500 gene mutations have been described in various types of malignancies, hematologic as well as solid tumors. Intact p53 function is essential for the maintenance of the non-tumorogenic phenotype of cells. Thus, p53 plays a vital role in suppressing the development of cancer.

Physical properties

GST-tagged, full-length human p53

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandra Ward et al.
BMC cancer, 11, 71-71 (2011-02-18)
Hepatocellular carcinoma (HCC), one of the most common cancers world-wide occurs twice as often in men compared to women. Predisposing conditions such as alcoholism, chronic viral hepatitis, aflatoxin B1 ingestion, and cirrhosis all contribute to the development of HCC. We
Dania Alarcon-Vargas et al.
Carcinogenesis, 23(4), 541-547 (2002-04-19)
The p53-Mdm2 paradigm represents the best-studied relationship between a tumor suppressor gene which functions as a transcription factor and an oncogene, which functions primarily as an E3 protein ligase. The intimate relationship between these two partners has expanded to include
Se Kyung Lee et al.
PloS one, 10(8), e0124658-e0124658 (2015-08-05)
Overexpression of p53 is the most frequent genetic alteration in breast cancer. Recently, many studies have shown that the expression of mutant p53 differs for each subtype of breast cancer and is associated with different prognoses. In this study, we
Cristina Muñoz-San Martín et al.
Analytical and bioanalytical chemistry, 412(24), 6177-6188 (2020-01-29)
Proteases are involved in cancer' taking part in immune (dis)regulation, malignant progression and tumour growth. Recently, it has been found that expression levels of one of the members of the serine protease family, trypsin, is upregulated in human cancer cells
Brandon S Sheffield et al.
The American journal of surgical pathology, 39(7), 977-982 (2015-01-31)
A variety of immunohistochemical (IHC) stains have been proposed to mark either benign or malignant mesothelial proliferations. Loss of the p16 tumor suppressor (CDKN2A), through homozygous deletions of 9p21, is a good marker of mesotheliomas but lacks sensitivity. Recent reports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service