Skip to Content
Merck
All Photos(4)

Key Documents

L9150

Sigma-Aldrich

N-Lauroylsarcosine sodium salt

detergent for use in cell lysis

Synonym(s):

Sodium lauroyl sarcosinate, N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Beilstein:
5322974
EC Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

description

anionic

product line

BioReagent

Assay

≥94%

form

powder

mol wt

micellar avg mol wt 600

aggregation number

2

technique(s)

protein purification: suitable
protein quantification: suitable

CMC

14.6 mM (20-25°C)

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

foreign activity

DNase, RNase, none detected

storage temp.

room temp

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium lauroyl sarcosinate, the sodium salt of an acyl derivative of sarcosine—a natural amino acid occurring in muscles and body tissues—stands out as a valuable component in molecular biology, biochemical, and cell biology research. Derived from a natural fatty acid and the amino acid derivative sarcosine, N-Lauroylsarcosine is a biodegradable anionic surfactant usually synthesized through acylation and neutralization reactions involving lauric acid and sodium sarcosine.

This amino acid-type anionic surfactant, also recognized as a fatty acid amide and detergent, exhibits protein denaturant potency and has been integrated into lysing solutions, such as the cornet assay. Its application extends to lysing protoplasts for chromosomal DNA isolation from Streptococcus mutans. Notably, Lauroylsarcosine Sodium Salt, characterized by its ionic detergent properties, is commonly employed for purifying membrane proteins. Additionally, Sodium Lauroyl Sarcosine also plays a crucial role in concentrated salt solutions, facilitating cell lysis in RNA purification protocols and membrane solubilization. Whether it′s breaking down cell structures or facilitating membrane protein purification, this mild surfactant proves valuable in various applications within the realms of biology and biochemistry.

Application

Suitable for
  • solubilization and separation of membrane proteins
  • lysis of cells during the isolation of RNA
  • inhibition of hexokinase

Features and Benefits

  • Suitable for Molecular Biology, Biochemical and Cell Biology research
  • Can be used for Protein Purification and Quantification
  • High purity product for research applications

Other Notes

Anionic surfactant.
For additional information on our range of Biochemicals, please complete this form.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

512.6 °F - closed cup

Flash Point(C)

267 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Swenshon et al.
Journal of clinical microbiology, 36(7), 1902-1906 (1998-07-03)
The present study was designed to identify and comparatively investigate 35 beta-hemolytic streptococci isolated from stranded harbor porpoises or from animals caught in fishing nets of the North and Baltic seas. According to biochemical and serological data and to lectin
A R Günthert et al.
The Journal of cell biology, 134(4), 1089-1096 (1996-08-01)
Ligation of CD95 (APO-1/Fas) cell surface receptors induces death in apoptosis-sensitive cells. Induction of apoptosis in adherent gamma interferon-stimulated HT-29 and COLO 205 colon carcinoma cells by cross-linking CD95 with anti-APO-1 monoclonal antibody resulted in detachment of the cells from
Rhonda I Hobb et al.
Microbiology (Reading, England), 155(Pt 3), 979-988 (2009-02-28)
Although infection with Campylobacter jejuni is one of the leading causes of gastroenteritis worldwide, relatively little is known about the factors that are required to elicit a protective immune response. The need for a vaccine against this pathogen is well
Joanna A Korecka et al.
PloS one, 8(5), e63862-e63862 (2013-06-01)
Multiple genetic and environmental factors play a role in the development and progression of Parkinson's disease (PD). The main neuropathological hallmark of PD is the degeneration of dopaminergic (DAergic) neurons in the substantia nigra pars compacta. To study genetic and
Nadine Schrode et al.
Nature genetics, 51(10), 1475-1485 (2019-09-25)
The mechanisms by which common risk variants of small effect interact to contribute to complex genetic disorders are unclear. Here, we apply a genetic approach, using isogenic human induced pluripotent stem cells, to evaluate the effects of schizophrenia (SZ)-associated common

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service