Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
-Lauroylsarcosine sodium salt
Synonym(s):
N
-Dodecanoyl-
N
-methylglycine sodium salt, Sarkosyl NL
Linear Formula:
CH
3
(CH
2
)
10
CON(CH
3
)CH
2
COONa
CAS Number:
137-16-6
Molecular Weight:
293.38
EC Number:
205-281-5
Products
Product Category
detergents (5)
Brand
Sigma-Aldrich (5)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
L9150
N
-Lauroylsarcosine sodium salt,
detergent for use in cell lysis
Expand
View Pricing
L5125
N
-Lauroylsarcosine sodium salt,
≥94%
Expand
View Pricing
61743
N
-Lauroylsarcosine sodium salt,
BioUltra, for molecular biology, ≥99.0% (HPLC)
Expand
View Pricing
61745
N
-Lauroylsarcosine sodium salt,
≥97.0% (HPLC)
Expand
View Pricing
L5777
N
-Lauroylsarcosine sodium salt,
BioXtra, ≥97% (TLC)
Expand
View Pricing