Skip to Content
Merck
All Photos(1)

Key Documents

L7007

Sigma-Aldrich

Lysergic acid diethylamide

Synonym(s):

LSD, Lysergide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N3O
CAS Number:
Molecular Weight:
323.43
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Quality Level

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

storage temp.

−20°C

SMILES string

[H][C@@]12Cc3c[nH]c4cccc(C1=C[C@H](CN2C)C(=O)N(CC)CC)c34

InChI

1S/C20H25N3O/c1-4-23(5-2)20(24)14-9-16-15-7-6-8-17-19(15)13(11-21-17)10-18(16)22(3)12-14/h6-9,11,14,18,21H,4-5,10,12H2,1-3H3/t14-,18-/m1/s1

InChI key

VAYOSLLFUXYJDT-RDTXWAMCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent hallucinogen; non-selective serotonin receptor agonist.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ingrid Garbus et al.
PloS one, 12(11), e0185595-e0185595 (2017-11-02)
A long-standing goal in plant breeding has been the ability to confer apomixis to agriculturally relevant species, which would require a deeper comprehension of the molecular basis of apomictic regulatory mechanisms. Eragrostis curvula (Schrad.) Nees is a perennial grass that
Irene Donati et al.
Horticulture research, 5, 56-56 (2018-11-06)
Flowers can provide a protected and nutrient-rich environment to the epiphytic microflora, thus representing a sensible entry point for pathogens such as Pseudomonas syringae pv. actinidiae (Psa). This bacterium can colonize both male and female Actinidia flowers, causing flower browning
Differential Classical Conditioning of the Nocebo Effect: Increasing Heat-Pain Perception without Verbal Suggestions.
Anne-Kathrin Bräscher et al.
Frontiers in psychology, 8, 2163-2163 (2018-01-13)
Mohammad B Habibi Najafi et al.
Food science & nutrition, 7(8), 2684-2691 (2019-08-21)
Instrumental hardness and sensory characteristics of pistachio nuts such as flavor, texture, and overall acceptability were measured in three different packages (atmosphere, vacuum, and oxygen scavenger), at three different conditions temperatures (20, 35, and 50˚C) during 12 weeks' storage. Results showed
Mara Vegro et al.
Journal of agricultural and food chemistry, 64(48), 9224-9236 (2016-12-10)
Freshly consumed apples (Malus domestica L. Borkh) can cause allergic reactions because of the presence of four classes of allergens. Knowledge of the genetic factors affecting the allergenic potential of apples would provide important information for the selection of hypoallergenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service