Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

Merck

Key Documents

Skip To

C5776

Cocaine hydrochloride

CNS stimulator, powder

Synonym(s):

Ecgonine methyl ester benzoate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · HCl
CAS Number:
Molecular Weight:
339.81
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-167-1
MDL number:
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cocaine hydrochloride,

form

powder

Quality Segment

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal), kontrollierte Droge in Deutschland

application(s)

forensics and toxicology

SMILES string

Cl.COC(=O)[C@H]1[C@H](C[C@@H]2CC[C@H]1N2C)OC(=O)c3ccccc3

InChI

1S/C17H21NO4.ClH/c1-18-12-8-9-13(18)15(17(20)21-2)14(10-12)22-16(19)11-6-4-3-5-7-11;/h3-7,12-15H,8-10H2,1-2H3;1H/t12-,13+,14-,15+;/m0./s1

InChI key

PIQVDUKEQYOJNR-VZXSFKIWSA-N

Application

Cocaine hydrochloride has been used:
  • in saline control to study the effects of cocaine self-administration in mouse brain
  • as adopamine (DA), noradrenaline (NA), and serotonin (5-HT) reuptake inhibitor to evoke neurotransmitter release in artificial fish cerebrospinal fluid (aCSF)
  • to examine conditioning to intravenous cocaine hydrochloride

Biochem/physiol Actions

Inhibits the dopamine, norepinephrine, and serotonin transporters with Kis of about 300 nM, 900 nM, and 400 nM, respectively. Unlike amphetamines, it has no effect on catecholamine release.
Inhibits the dopamine, norepinephrine, and serotonin transporters.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C8912M0267C222
description

-

description

-

description

powder, ≥98%

description

-

form

powder

form

powder

form

powder

form

powder

drug control

kontrollierte Droge in Deutschland, USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal); Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

-

application(s)

forensics and toxicology


Still not finding the right product?

Explore all of our products under Cocaine hydrochloride


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Repr. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. I wonder that if I dissolve the cocaine in saline, is it stable for long term? Should I storage it in -20 or 4? For how long?

    1 answer
    1. This product is sold as a white, photosensitive solid. The data sheet indicates solubility is 2.5 g/mL in water or 300 mg/mL in ethanol. No information is available regarding solution stability after solubilizing the cocaine. A certified reference standard is available as product C1528, with a concentration of 1 mg/mL in methanol. The recommended storage temperature for this product is 2-8 °C. A random Certificate of Analysis shows the product has a 3-year expiration date from the date of quality release.

      Helpful?

Reviews

No rating value

Active Filters