Skip to Content
Merck

L6150

Sigma-Aldrich

Lysine Oxidase from Trichoderma viride

lyophilized powder, ≥20 units/mg protein

Synonym(s):

L-Lysine:oxygen oxidoreductase (deaminating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Pricing and availability is not currently available.

biological source

fungus (Trichoderma viride)

Quality Level

form

lyophilized powder

specific activity

≥20 units/mg protein

mol wt

112 kDa

composition

Protein, 5-20%

storage temp.

2-8°C

General description

Lysine Oxidase from Trichoderma viride is a homodimeric flavoenzyme corresponding to molecular mass of 112 kDa.[1] It is stable at 65°C and is highly specific for L-lysine substrate.[1] It comprises FAD-binding, substrate binding and a helical domain with distinct active site funnel.[1]

Application

Lysine Oxidase from Trichoderma viride has been used in the preparation of luminescent biochip preparation.[2]

Biochem/physiol Actions

Lysine Oxidase from Trichoderma viride catalyzes the formation of α-keto- ε-aminocaproate by the oxidative deamination of L-lysine. It displays anti-tumor functionality in cancer leukaemic cells.[1] It is a tumor suppressor for squamous cell, fibroblast, ovarian and gastric tumors.[3] Lysine oxidase also plays key role in connective tissue structural integrity and embryo development.[3]

Unit Definition

One unit will catalyze the formation of 1 μmole of 6-amino-2-oxohexanoic acid from L-lysine per min at 37°C at pH 8.0.

Physical form

Contains phosphate buffer salts and stabilizer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I P Smirnova et al.
Voprosy meditsinskoi khimii, 46(4), 384-387 (2000-11-15)
The ability of protein isolated from (Trichoderma Rifai) and azydothymidine to inhibit the reproduction of HIV-virus was compared. The obtained experimental data have verified that Trichoderma Rifai protein is a promising human immunodeficiency virus (HIV) inhibitor.
Seiji Okazaki et al.
Journal of biochemistry, 154(3), 233-236 (2013-08-03)
We have determined the x-ray crystal structure of L-lysine ε-oxidase from Marinomonas mediterranea in its native and L-lysine-complex forms at 1.94- and 1.99-Å resolution, respectively. In the native enzyme, electron densities clearly indicate the presence of cysteine tryptophylquinone (CTQ) previously
E V Lukasheva et al.
Biochemistry. Biokhimiia, 67(10), 1152-1158 (2002-12-04)
This review summarizes data on the properties of L-lysine alpha-oxidase, an enzyme that belongs to the group of oxidases of L-amino acids. This enzyme acts virtually only on L-lysine with a rather low Km yielding alpha-keto-epsilon-aminocaproic acid. The decrease in
E E Ferapontova et al.
Biochemistry. Biokhimiia, 66(8), 832-839 (2001-09-22)
Adsorption and bioelectrocatalytic activity of native horseradish peroxidase (HRP) and its recombinant forms on polycrystalline gold electrodes were studied. Recombinant forms of HRP were produced by a genetic engineering approach using an E. coli expression system. According to direct mass
Vadim S Pokrovsky et al.
Anti-cancer drugs, 24(8), 846-851 (2013-06-20)
L-Lysine α-oxidase (LO) from a novel Trichoderma strain: Trichoderma cf. aureoviride Rifai shows favorable biochemical and kinetic properties (Km for L-lysine of 17.9 µmol/l, optimum pH 8.0, high stability) and significant antiproliferative activity both in vitro and in vivo. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service