Skip to Content
Merck
All Photos(1)

Key Documents

L5502

Sigma-Aldrich

Lys-Lys dihydrochloride

≥98% (TLC)

Synonym(s):

Dilysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H26N4O3 · 2HCl
CAS Number:
Molecular Weight:
347.28
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Lys-Lys dihydrochloride,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

Cl.NCCCCC(N)C(=O)NC(CCCCN)C(O)=O

InChI

1S/C12H26N4O3.ClH/c13-7-3-1-5-9(15)11(17)16-10(12(18)19)6-2-4-8-14;/h9-10H,1-8,13-15H2,(H,16,17)(H,18,19);1H

InChI key

ROGKVZGKYWMMAT-UHFFFAOYSA-N

Application

Lysyllysine (Lys-Lys) may be used in studies of prebiotically relevant Salt-Induced Peptide Formation (SIPF) and in for physical chemical analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R T MacGillivray et al.
Biochemistry, 39(6), 1211-1216 (2000-02-24)
Serum transferrin is the major iron transport protein in humans. Its function depends on its ability to bind iron with very high affinity, yet to release this bound iron at the lower intracellular pH. Possible explanations for the release of
Heather M Baker et al.
Acta crystallographica. Section D, Biological crystallography, 63(Pt 3), 408-414 (2007-03-01)
Iron uptake by humans depends on the ability of the serum protein transferrin (Tf) to bind iron as Fe(3+) with high affinity but reversibly. Iron release into cells occurs through receptor-mediated endocytosis, aided by the lower endosomal pH of about
Roland Kabuss et al.
Glycobiology, 15(10), 905-911 (2005-06-04)
Nucleotide-sugar transporters supply mainly the Golgi glycosyltransferases with substrates. Some glycosyltransferases in the endoplasmic reticulum (ER), however, also use activated sugars. Recent studies have demonstrated that UDP-galactose (UDP-Gal) is the substrate for the ER resident ceramide-galactosyltransferase (cer-GalT) and cells expressing
Lauren P Jackson et al.
Developmental cell, 23(6), 1255-1262 (2012-11-28)
COPI mediates retrograde trafficking from the Golgi to the endoplasmic reticulum (ER) and within the Golgi stack, sorting transmembrane proteins bearing C-terminal KKxx or KxKxx motifs. The structure of KxKxx motifs bound to the N-terminal WD-repeat domain of β'-COP identifies
Erik Lontok et al.
Journal of virology, 78(11), 5913-5922 (2004-05-14)
Coronavirus budding at the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) requires accumulation of the viral envelope proteins at this point in the secretory pathway. Here we demonstrate that the spike (S) protein from the group 3 coronavirus infectious bronchitis virus (IBV)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service