Skip to Content
Merck
All Photos(1)

Key Documents

G3640

Sigma-Aldrich

Glu-Glu

≥98% (TLC)

Synonym(s):

L-α-glutamyl-L-glutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O7
CAS Number:
Molecular Weight:
276.24
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Glu-Glu,

Assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(O)=O

InChI

1S/C10H16N2O7/c11-5(1-3-7(13)14)9(17)12-6(10(18)19)2-4-8(15)16/h5-6H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)

InChI key

KOSRFJWDECSPRO-UHFFFAOYSA-N

Biochem/physiol Actions

Glu-Glu is a small anionic dipeptides that may be used in physicochemical analysis of conformations states and protonation patterns.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marieke Schor et al.
Physical chemistry chemical physics : PCCP, 13(22), 10457-10467 (2011-04-20)
Triblock copolymers consisting of a silk-based ((Gly-Ala)(3)Gly-Glu) repeat flanked by hydrophilic outer blocks self-assemble into micrometer long fibrils in response to a trigger. Since the exact mechanism of the fibril formation remains unclear, we employ a multiscale modelling approach in
Sophie Laborie et al.
JPEN. Journal of parenteral and enteral nutrition, 39(6), 729-737 (2014-06-14)
Oxidant stress is implicated in the pathogenesis of bronchopulmonary dysplasia (BPD). Light induces peroxide generation in parenteral nutrition (PN) solutions, creating an oxidant stress. Shielding PN from light decreases its peroxide content, which has nutrition and biochemical benefits in animals
Muhammad A Hagras et al.
The journal of physical chemistry. B, 119(46), 14637-14651 (2015-10-28)
The most detailed and comprehensive to date study of electron transfer reactions in the respiratory complex III of aerobic cells, also known as bc1 complex, is reported. In the framework of the tunneling current theory, electron tunneling rates and atomistic
C J Jeon et al.
Visual neuroscience, 14(1), 27-37 (1997-01-01)
Biochemical studies provide evidence that the pathway from visual cortex to the superior colliculus (SC) utilizes glutamate as a neurotransmitter. In the present study, we have used immunocytochemistry, visual cortex lesions, and retrograde tracing to show directly by anatomical methods
Nobuhiro Takahashi et al.
Oral microbiology and immunology, 17(1), 50-54 (2002-02-28)
Porphyromonas gingivalis, Prevotella intermedia, Prevotella nigrescens and Fusobacterium nucleatum, which can frequently be isolated from periodontal pockets, preferentially utilize proteins and peptides as growth substrates. In this study, we determined the size of peptide that is preferentially utilized as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service