Skip to Content
Merck
All Photos(1)

Key Documents

L135

Sigma-Aldrich

L-741,626

≥98% (HPLC)

Synonym(s):

(±)-3-[4-(4-Chlorophenyl)-4-hydroxypiperidinyl]methylindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21ClN2O
CAS Number:
Molecular Weight:
340.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥20 mg/mL
ethanol: 24 mg/mL
H2O: insoluble

storage temp.

room temp

SMILES string

OC1(CCN(CC1)Cc2c[nH]c3ccccc23)c4ccc(Cl)cc4

InChI

1S/C20H21ClN2O/c21-17-7-5-16(6-8-17)20(24)9-11-23(12-10-20)14-15-13-22-19-4-2-1-3-18(15)19/h1-8,13,22,24H,9-12,14H2

InChI key

LLBLNMUONVVVPG-UHFFFAOYSA-N

Gene Information

Application

L-741,626 has been used as selective dopamine (D2) receptor antagonist to study its effects on the uptake of dopamine in human embryonic kidney 293 (HEK293) cells. It has also been used as a D2 receptor antagonist to study its effects on thyrotropin-releasing hormone (TRH) induced prolactin release.

Biochem/physiol Actions

L-741,626 is a selective dopamine (D2) receptor antagonist.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B J Bowery et al.
British journal of pharmacology, 119(7), 1491-1497 (1996-12-01)
1. The ability of PD 128907 to activate dopamine receptors in the ventral tegmental area, substantia nigra pars compacta, and striatum was investigated by use of in vitro electrophysiological recording and fast cyclic voltammetry. The affinity of a novel D2
Alexander Gont et al.
Oncotarget, 8(5), 8559-8573 (2017-01-05)
A defining feature of the brain cancer glioblastoma is its highly invasive nature. When glioblastoma cells are isolated from patients using serum free conditions, they accurately recapitulate this invasive behaviour in animal models. The Rac subclass of Rho GTPases has
Masaki Nakano et al.
General and comparative endocrinology, 168(2), 287-292 (2010-06-18)
Dopamine receptors in mammals are known to consist of two D1-like receptors (D1 and D5) and three D2-like receptors (D2, D3 and D4). The aim of this study was to determine the dopamine receptor subtype that mediates the inhibitory action
Florence Loiseau et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 19(1), 23-33 (2008-09-17)
Though D(3) receptor antagonists can enhance cognitive function, their sites of action remain unexplored. This issue was addressed employing a model of social recognition in rats, and the actions of D(3) antagonists were compared to D(1) agonists that likewise possess
Ferran Jardí et al.
Scientific reports, 8(1), 957-957 (2018-01-19)
Low testosterone (T) in men, especially its free fraction, has been associated with loss of energy. In accordance, orchidectomy (ORX) in rodents results in decreased physical activity. Still, the mechanisms through which T stimulates activity remain mostly obscure. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service