Skip to Content
Merck
All Photos(2)

Documents

I5281

SAFC

L-Isoleucine

Synonym(s):

(2S,3S)-2-Amino-3-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721792
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
NACRES:
NA.26

biological source

non-animal source

Assay

≥98.5%

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, heavy metals, residual solvents, tested

mp

288 °C (dec.) (lit.)

solubility

1 M HCl: 50 mg/mL

application(s)

pharmaceutical (small molecule)

foreign activity

cytotoxicity, tested

SMILES string

CC[C@H](C)[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1

InChI key

AGPKZVBTJJNPAG-WHFBIAKZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to the documentation for this product requires a confidentiality disclosure agreement.

Application

L-Isoleucine is an essential amino acid. It can be biosynthesized from threonine or through fermentation of carbohydrates. The amino acid L-isoleucine is an isomer of L-Leucine, differing in the position of the side chain. It is also considered to be a branched chain amino acid, along with L-Leucine and L-Valine, which are sources of fuel and/or transport during cell development. It is used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eva Blomstrand et al.
The Journal of nutrition, 136(1 Suppl), 269S-273S (2005-12-21)
BCAAs (leucine, isoleucine, and valine), particularly leucine, have anabolic effects on protein metabolism by increasing the rate of protein synthesis and decreasing the rate of protein degradation in resting human muscle. Also, during recovery from endurance exercise, BCAAs were found
L Edvinsson
Blood vessels, 28(1-3), 35-45 (1991-01-01)
The cerebral circulation is supplied with two vasodilator systems: the parasympathetic system storing vasoactive intestinal peptide, peptide histidine isoleucine, acetylcholine and in a subpopulation of nerves neuropeptide Y, and the sensory system, mainly originating in the trigeminal ganglion, storing substance
Toshihiro Sato et al.
The Journal of biological chemistry, 278(6), 4314-4321 (2002-11-22)
Previously, we determined the crystal structures of the dimeric ligand binding region of the metabotropic glutamate receptor subtype 1. Each protomer binds l-glutamate within the crevice between the LB1 and LB2 domains. We proposed that the two different conformations of
Urho M Kujala et al.
Circulation, 127(3), 340-348 (2012-12-22)
Long-term physical inactivity seems to cause many health problems. We studied whether persistent physical activity compared with inactivity has a global effect on serum metabolome toward reduced cardiometabolic disease risk. Sixteen same-sex twin pairs (mean age, 60 years) were selected
Adeline Chauvin et al.
The New phytologist, 197(2), 566-575 (2012-11-23)
Damage-inducible defenses in plants are controlled in part by jasmonates, fatty acid-derived regulators that start to accumulate within 30 s of wounding a leaf. Using liquid chromatography-tandem mass spectrometry, we sought to identify the 13-lipoxygenases (13-LOXs) that initiate wound-induced jasmonate synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service