Skip to Content
Merck
All Photos(1)

Key Documents

I7160

Sigma-Aldrich

IMS2186

≥98% (HPLC), solid

Synonym(s):

3-[1-(3-Hydroxy-4-methoxyphenyl)-meth-(E)-ylidene]-6-methyl-chroman-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O4
CAS Number:
Molecular Weight:
296.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

COc1ccc(cc1O)\C=C2/COc3ccc(C)cc3C2=O

InChI

1S/C18H16O4/c1-11-3-5-16-14(7-11)18(20)13(10-22-16)8-12-4-6-17(21-2)15(19)9-12/h3-9,19H,10H2,1-2H3/b13-8+

InChI key

BZCADTHMZLYKAR-MDWZMJQESA-N

Biochem/physiol Actions

IMS2186 is an anti-proliferative and anti-angiogenic agent. Apparently IMS2186 blocks the cell cycle at G2 and inhibits the production of PGE2/TNF-α. IMS2186 inhibits cell growth in vitro in tumor cells, non-transformed fibroblasts, and retinal pigment epithelial cells. IMS2186 inhibits angiogenesis (IC50 = 0.1 μM) and cell migration. Intraocular injection of IMS2186 could be a long-lasting effective treatment for CNV (choroidal neovascularization) in AMD (age-related macular degeneration) with a reduction in scarring and related visual loss.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service