Skip to Content
Merck
All Photos(1)

Key Documents

I4032

Sigma-Aldrich

Interleukin-21 from mouse

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IL-21

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

40-120 ng/mL ED50

mol wt

predicted mol wt ~14.4 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il21(60505)

Biochem/physiol Actions

IL-21 (interleukin-21) belongs to the IL-2 cytokine family. It is secreted by CD4+ T, natural killer (NK) and Th17 cells. IL-21 associates with IL-21 receptor and participates in B-cell proliferation, differentiation as well as apoptosis. It also promotes the cytotoxic activity of CD8+ T cells and natural killer cells.IL-21 plays a crucial role in the differentiation of follicular helper T cells. It also exhibits antitumor activity. It is also involved in the activation of JAK-STAT (Janus kinase - signal transducer and activator of transcription), PI3K (phosphatidylinositol 3-kinase) and MAPK (mitogen activated protein kinase) pathways.
IL-21 and its receptor have important roles in T and B cell proliferation and natural killer cell maturation. Human and mouse IL-21 share 57% amino acid identity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to induce proliferation of human N1186 T cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo D Becker et al.
PloS one, 5(10) (2010-10-20)
Passive transfer of antibodies not only provides immediate short-term protection against disease, but also can be exploited as a therapeutic tool. However, the 'humanization' of murine monoclonal antibodies (mAbs) is a time-consuming and expensive process that has the inherent drawback
Shruti Bhatt et al.
Blood, 126(13), 1555-1564 (2015-07-22)
Mantle cell lymphoma (MCL) is a distinct subtype of non-Hodgkin lymphoma characterized by overexpression of cyclin D1 in 95% of patients. MCL patients experience frequent relapses resulting in median survival of 3 to 5 years, requiring more efficient therapeutic regimens.
Taishi Takahara et al.
Cancer science, 107(11), 1572-1580 (2016-08-26)
Diffuse large B-cell lymphoma (DLBCL) is the most common subtype of malignant lymphoma; it derives from germinal center B cells. Although DLBCL harbors many genetic alterations, synergistic roles between such alterations in the development of lymphoma are largely undefined. We
S Lajoie et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 44(7), 976-985 (2014-05-09)
Interleukin-21 (IL-21) has been implicated in the development of Th2-mediated immune responses; however, the exact role it plays in allergic diseases is not well understood. To elucidate the contribution of IL-21 receptor signalling to Th2-dependent immune responses in the lung.
Mark J Kwakkenbos et al.
Nature medicine, 16(1), 123-128 (2009-12-22)
The B cell lymphoma-6 (Bcl-6) and Bcl-xL proteins are expressed in germinal center B cells and enable them to endure the proliferative and mutagenic environment of the germinal center. By introducing these genes into peripheral blood memory B cells and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service