Skip to Content
Merck
All Photos(1)

Key Documents

MAB3300

Sigma-Aldrich

Anti-TIMP-1 Antibody, clone 7-6C1

clone 7-6C1, Chemicon®, from mouse

Synonym(s):

Tissue Inhibitor of Metalloproteinase-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7-6C1, monoclonal

species reactivity

rat, human, bovine

should not react with

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TIMP1(7076)

General description

Tissue Inhibitors of Metalloproteinases (TIMPs) inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells. TIMP-1 and TIMP-2 have similar properties, specifically in inhibiting enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-1 forms a complex of 1:1 stoichiometry with activated interstitial collagenase, activated stromelysin, active form of 72 kDa type IV collagenase (also known as MMP-2 or gelatinase A), and latent and active forms 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-1 levels in quiescent cells and tissues is low (pg/ml), and stimulation with the phorbol ester TPA or protein concentration is often needed to visualize protein bands. In addition, cell types differ greatly in the amount of TIMP-1 produced.

Specificity

MAB3300 is a purified mouse monoclonal antibody to bovine tissue inhibitor of metalloproteinases-1 (bovine TIMP-1). The antibody specifically reacts with bovine TIMP-1 and cross-reacts with human and rat TIMP-1, but not with mouse TIMP-1. By western blot the antibody recognizes a glycoprotein of 28.5 kDa, which is identified as tissue inhibitor of metalloproteinases-1 (TIMP-1).

Immunogen

Bovine TIMP-1

Application

Detect TIMP-1 using this Anti-TIMP-1 Antibody, clone 7-6C1 validated for use in ELISA, WB, IH.
Immunoblotting: 1:1000, identifies a band of approximately 30KDa. Note TIMP-1 is found in very low concentrations in most cells and tissues.Positive control is HT1080 cells treated with PMA or recombinant TIMP-1 protein CC3328.

Immunohistochemistry on frozen sections or acetone fixed tissues. Formalin fixed paraffin-embedded tissue sections are usually not reactive. PLP fixation is suggested as an alternative.

EIA

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs

Linkage

Replaces: 04-1133

Physical form

Format: Purified
Purified immunoglobulin presented as a liquid in 0.1 M Na-Phosphate buffer, pH 7.0 containing protease free bovine serum albumin.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze-thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lawrence D True et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 23(10), 1346-1356 (2010-06-22)
A by-product in the processing of prostate tissue for cell sorting by collagenase digestion is the media supernatant that remains after the cells are harvested. These supernatants contain proteins made by the cells within the tissue. Quantitative proteomic analysis of
Weiheng Wang et al.
International journal of molecular medicine, 41(5), 2553-2564 (2018-02-14)
Transplantation of nucleus pulposus cells (NPCs) into the intervertebral disc (IVD) has been demonstrated to be an effective treatment of degenerative disc disease (DDD). However, the underlying mechanisms have remained to be sufficiently elucidated. The aim of the present study
Rhonda Souvenir et al.
Neurobiology of disease, 44(1), 28-37 (2011-06-22)
Previous studies have shown that erythropoietin (EPO) is neuroprotective in both in vivo and in vitro models of hypoxia ischemia. However these studies hold limited clinical translations because the underlying mechanism remains unclear and the key molecules involved in EPO-induced
Jia-Hau Yen et al.
Molecules (Basel, Switzerland), 27(5) (2022-03-11)
This study aims to investigate the wound-healing effectiveness of the phenolic compound, naringin, both in vitro and in vivo. Male mice were shaved on their dorsal skin under isoflurane, a biopsy punch was made in four symmetrical circular resection windows
Felix Wittig et al.
Cells, 12(13) (2023-07-14)
Despite the well-described anticarcinogenic effects of endocannabinoids, the influence of the endocannabinoid system on tumor angiogenesis is still debated. In the present study, conditioned medium (CM) from A549 and H358 lung cancer cells treated with ascending concentrations of the monoacylglycerol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service