Skip to Content
Merck
All Photos(1)

Key Documents

I2633

Sigma-Aldrich

Cardiogreen

Powder

Synonym(s):

Foxgreen, IC Green, Indocyanine green

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H47N2NaO6S2
CAS Number:
Molecular Weight:
774.96
Beilstein:
4115884
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Cardiogreen, polymethine dye

form

powder

composition

Dye content, ≥75%

technique(s)

titration: suitable

color

green to very dark green

solubility

water: 0.01 g/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CC1(C)/C(=C\C=C\C=C\C=C\C2=[N+](CCCCS([O-])(=O)=O)c3ccc4ccccc4c3C2(C)C)N(CCCCS([O-])(=O)=O)c5ccc6ccccc6c15

InChI

1S/C43H48N2O6S2.Na/c1-42(2)38(44(28-14-16-30-52(46,47)48)36-26-24-32-18-10-12-20-34(32)40(36)42)22-8-6-5-7-9-23-39-43(3,4)41-35-21-13-11-19-33(35)25-27-37(41)45(39)29-15-17-31-53(49,50)51;/h5-13,18-27H,14-17,28-31H2,1-4H3,(H-,46,47,48,49,50,51);/q;+1/p-1

InChI key

MOFVSTNWEDAEEK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cardiogreen is a negatively charged polymethine dye that forms noncovalent fluorescent complexes with proteins. This "pseudofluorogenic" property has been utilized for protein determination by capillary electrophoresis with diode laser-induced fluorescence detection.

Application

Cardiogreen has been used to resolve several labeled proteins including: human serum albumin, ribonuclease A, transferrin, and cytochrome c.
Cardiogreen has been used as an intraocular capsular stain. It has been used in the dye-dilution method for cardiac output.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

solubility: 0.01g in 10mL H2O : green to very dark green

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M McCorquodale et al.
Electrophoresis, 22(12), 2403-2408 (2001-08-25)
Indocyanine green (ICG)--a negatively charged, polymethine dye--can interact noncovalently with proteins to form fluorescent complexes, with excitation and emission maxima near 780 and 820 nm, respectively. This behavior was realized utilizing either a 100 mM phosphate buffer or a 25
Mixed S-nitrosylated polymerized bovine hemoglobin species moderate hemodynamic effects in acutely hypoxic rats.
Irwin D et al.
American Journal of Respiratory Cell and Molecular Biology, 42, 200-200 (2010)
Aqueous Angiography with Fluorescein and Indocyanine Green in Bovine Eyes.
Huang AS et al.
Translational vision science & technology, 5, 5-5 (2016)
Toshiki Endo et al.
Journal of neurosurgery. Spine, 18(5), 443-449 (2013-03-12)
The characteristics and efficacy of indocyanine green (ICG) videoangiography in cavernous malformation (CM) have not been fully elucidated. The purpose of this paper is to examine the potential utility of ICG videoangiography in the surgical treatment of intramedullary CMs. The
Jun Shibasaki et al.
The Journal of pediatrics, 164(5), 1116-1120 (2014-02-13)
To investigate the use of indocyanine green (ICG) lymphography in the diagnosis and assessment of the severity of lymphatic dysfunction in infants and neonates with congenital lymphatic pleural effusion and ascites. We performed ICG lymphography on 10 neonates and infants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service