I1786
ID-8
≥98% (HPLC)
Synonym(s):
1-(4-Methoxyphenyl)-2-methyl-3-nitro-1H-indol-6-ol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H14N2O4
CAS Number:
Molecular Weight:
298.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
yellow
solubility
DMSO: ≥10 mg/mL
storage temp.
2-8°C
SMILES string
COc1ccc(cc1)-n2c(C)c([N+]([O-])=O)c3ccc(O)cc23
InChI
1S/C16H14N2O4/c1-10-16(18(20)21)14-8-5-12(19)9-15(14)17(10)11-3-6-13(22-2)7-4-11/h3-9,19H,1-2H3
InChI key
VVZNWYXIOADGSW-UHFFFAOYSA-N
General description
ID-8 is an inhibitor of dual-specificity tyrosine-phosphorylation-regulated kinase 1A (DYRK1A).
Biochem/physiol Actions
ID-8 sustains self-renewal and pluripotency of mouse embryonic stem cells (ESCs) in vitro. Stimulates proliferation at a steady rate (observed in serum-free media supplemented with 10 μM over a 30 day period).
Sustains self-renewal and pluripotency of mouse embryonic stem cells (ESCs) in vitro.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yusuke Higuchi et al.
Current molecular pharmacology, 9(3), 272-279 (2015-05-27)
Despite their high degree of identity and even higher homology, the two Kat3 transcriptional coactivators, CBP and p300, have distinct functions, particularly within the Wnt/β-catenin signaling cascade. ICG-001, by directly binding to CBP but not p300, inhibits CBP/β-catenin transcription and
Stefan Hindel et al.
PloS one, 10(6), e0128060-e0128060 (2015-06-11)
The purpose of our study was to validate perfusion quantification in a low-perfused tissue by dynamic contrast-enhanced magnetic resonance imaging (DCE-MRI) with shared k-space sampling using a blood pool contrast agent. Perfusion measurements were performed in a total of seven
A High-Throughput Screen Identifies DYRK1A inhibitor ID-8 that Stimulates Human Kidney Tubular Epithelial Cell Proliferation
Monteiro M B, et al.
Journal of the American Society of Nephrology, ASN-2018040392 (2018)
Jin Zhou et al.
Annals of translational medicine, 8(12), 752-752 (2020-07-11)
Paclitaxel is a widely used clinical first line chemotherapy drug for ovarian carcinoma. Tanshinone I (Tan-I) is one of the vital fat-soluble components, which derived from Chinese herbal medicine, Salvia miltiorrhiza Bunge. Herein, we evaluated whether Tan-I could enhance the
Jin Zhou et al.
Annals of translational medicine, 8(20), 1295-1295 (2020-11-20)
In Chinese herbal medicine, Tanshinone IIA (Tan-IIA) is one of the main compounds extracted from Salvia miltiorrhiza Bunge. Tan-IIA has been demonstrated to inhibit the growth of various tumors. However, the detailed molecular and cellular mechanisms of the antitumor effect
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service