Skip to Content
Merck
All Photos(1)

Key Documents

I1010

Sigma-Aldrich

IgA from human colostrum

reagent grade, ≥95%, essentially salt-free, lyophilized powder

Synonym(s):

Human IgA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

conjugate

unconjugated

Quality Level

grade

reagent grade

Assay

≥95%

form

essentially salt-free, lyophilized powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

IgA antibody is secretory antibody and is present abundantly in mucous linings of gastrointestinal, respiratory and genitourinary tracts, tears, colostrum and saliva. Thus secretory IgA contributes to the humoral defense mechanism against the pathogens on mucosal surfaces[1]
Human IgA is purified from pooled normal human colostrum by fractionation and ion-exchange fractionation.

Application

Purified IgA may be used as a reference antigen, standard, blocking agent or coating protein in a variety of immunoassays including ELISA, dot immunobinding, immunoblotting, immunodiffusion and immunoelectrophoresis. Other applications include starting materials for the preparation of immunogens and solidphase immunoadsorbents. IgA from human colostrum was used as a standard in ELISA (1 mg/ml)[2][3] and gel electrophoresis.[4]

Reconstitution

Reconstitute with buffered saline (150 mM sodium chloride).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauri Vehkala et al.
Annals of medicine, 45(8), 501-510 (2013-10-18)
Obesity and type 2 diabetes (T2D) associate with increased oxidative stress. Malondialdehyde acetaldehyde (MAA) adducts have been suggested to be one of the antigenic epitopes in MDA-LDL responsible for the antibody recognition. Our aim was to investigate the associations between
Hannah Karen Mina Labayo et al.
EClinicalMedicine, 27, 100561-100561 (2020-10-13)
Norovirus (NV) causes acute gastroenteritis in infants. Humoral and fecal immunoglobulin A (IgA) responses have been correlated with protection against NV; however, the role of breast milk IgA against NV infection and associated diarrhea is still unknown. This study aimed
E Knop et al.
Investigative ophthalmology & visual science, 42(3), 566-574 (2001-02-27)
Mucosa-associated lymphoid tissue (MALT) specifically protects mucosal surfaces. In a previous study of the human conjunctiva, evidence was also found for the presence of MALT in the lacrimal sac. The present study, therefore, aims to investigate its morphology and topographical
P Kosuwon et al.
Beneficial microbes, 9(4), 541-552 (2018-04-11)
Little is known about the impact of nutrition on toddler gut microbiota. The plasticity of the toddler gut microbiota indicates that nutritional modulation beyond infancy could potentially impact its maturation. The objective of this study was to determine the effect
Erick Sánchez-Salguero et al.
Maternal health, neonatology and perinatology, 5, 9-9 (2019-06-18)
Colostrum is the primary source of maternal immunoglobulin A (IgA) for the newborn. IgA participates in protection and regulation mechanisms of the immune response at the neonate's mucosa. Several studies have evaluated infectious diseases and vaccine protocols effects during pregnancy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service