Skip to Content
Merck

Skip To

401799

Human IgM, Plasma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.77
UNSPSC Code:
12352202
Clone:
-
Species reactivity:
-
Application:
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Human plasma)

Quality Segment

assay

>95% (SDS-PAGE)

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgM

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Native IgM from human plasma.

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 200 mM NaCl, 50 mM Tris, pH 8.0.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).
Prepared from plasma shown to be nonreactive for HBsAg and for antibodies to HCV, HBc, HIV-1, and HIV-2 by certified tests.

Other Notes

Contains both kappa and lambda light chains. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
401098401104401116
biological source

(Human plasma)

biological source

(Human plasma)

biological source

-

biological source

(Human plasma)

shipped in

wet ice

shipped in

wet ice

shipped in

ambient

shipped in

ambient

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

isotype

IgM

isotype

IgA

isotype

-

isotype

IgG

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters