Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
solution
Quality Segment
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
application(s)
hematology
histology
storage temp.
room temp
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description - | description suitable for microscopy | description - | description - |
| application(s) hematology | application(s) - | application(s) hematology | application(s) hematology |
| form solution | form - | form solution | form solution |
| IVD for in vitro diagnostic use | IVD - | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use |
| Quality Level 500 | Quality Level 100 | Quality Level 500 | Quality Level 500 |
| storage temp. room temp | storage temp. room temp | storage temp. room temp | storage temp. room temp |
| shelf life Expiry date on the label. | shelf life limited shelf life, expiry date on the label | shelf life Expiry date on the label. | shelf life Expiry date on the label. |
Still not finding the right product?
Explore all of our products under Malachite Green Solution
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
8A - Combustible corrosive hazardous materials
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



