Skip to Content
Merck

HT8028

Malachite Green Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
41116121
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

hematology
histology
microbiology

storage temp.

room temp

Application

Intended for use with Carbol Fuchsin (HT80-1) in Sigma Acid Fast Stain, Procedure HT80. Acts as both a counterstain and a differentiator.

Other Notes

Malachite green oxalate (certified), 1.5%; acetic acid, 10%; and glycerol, 17%, in deionized water.


Still not finding the right product?

Explore all of our products under Malachite Green Solution


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

No data available

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library