Merck
Skip To
HS5360A
Benchtop Counter
pack of 1 ea
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS5360A
General description
The Mechanical Benchtop Counter is perfect for blood cell (label included), bacterial colonies or other repetitive counting.
Features and Benefits
  • 6-Key Benchtop Counter (each key records up to 999)
  • Total window keeps track of total number of strokes made on all other keys; bell rings when total reaches 100
  • Blood Cell Key labels included
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
HS5360BPHCC360250PHCC360500
description
pack of 1 ea
description
9 Key
description
60 μm sensors designed to count particles between 6 μm & 36 μm
description
60 μm sensors designed to count particles between 6 μm & 36 μm
manufacturer/tradename
Heathrow Scientific HS5360A
manufacturer/tradename
Heathrow Scientific
manufacturer/tradename
Scepter
manufacturer/tradename
Scepter
packaging
pack of 1 ea
packaging
-
packaging
pkg of 250 ea
packaging
pkg of 500 ea
Still not finding the right product?
Explore all of our products under Benchtop Counter
Choose from one of the most recent versions:
Certificates of Analysis (COA)