Skip to Content
Merck
All Photos(6)

Key Documents

HPA044186

Sigma-Aldrich

Anti-NUDT16L1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Nudix (Nucleoside Diphosphate Linked Moiety X)-Type Motif 16-Like 1, Anti-SDOS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AVPELKQISRVEAMRLGPGWSHSCHAMLYAANPGQLFGRIPMRFSVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Nudix hydrolase 16 like 1 (NUDT16L1) also known as TIRR (tudor interacting repair regulator), specifically interacts with the ionizing radiation induced foci formation (IRIF) region of P53 binding protein 1 (53BP1). The gene is located on human chromosome 16p13.3. The protein is a member of nucleoside diphosphate-linked moiety X (Nudix) hydrolases family.

Immunogen

nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-NUDT16L1 antibody produced in rabbit has been used in western blotting and immunoprecipitation.

Biochem/physiol Actions

Nudix hydrolase 16 like 1 (NUDT16L1) acts as an inhibitor of P53 binding protein 1 (53BP1). The protein repairs double strand breaks by masking the histone methyl-lysine binding function of P53 binding protein 1 (53BP1).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82303

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein Syndesmos is a novel RNA-binding protein that regulates primary cilia formation
Avolio R et al.
Nucleic Acids Research (2018)
The p53-binding protein 1-Tudor interacting repair regulator complex participates in the DNA damage response
Zhang A et al.
The Journal of Biological Chemistry, jbc-M117 (2017)
TIRR regulates 53BP1 by masking its histone methyl-lysine binding function
Drane P et al.
Nature, 543(7644), 211-211 (2017)
The Nudix hydrolase superfamily
McLennan, AG
Cellular and Molecular Life Sciences, 63(2), 123-143 (2006)
Nishita Parnandi et al.
Molecular cell, 81(12), 2583-2595 (2021-05-08)
53BP1 influences genome stability via two independent mechanisms: (1) regulating DNA double-strand break (DSB) repair and (2) enhancing p53 activity. We discovered a protein, Tudor-interacting repair regulator (TIRR), that associates with the 53BP1 Tudor domain and prevents its recruitment to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service