Skip to Content
Merck
All Photos(2)

Key Documents

05892791001

Roche

cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail

Tablets supplied in foil blister packs.

Synonym(s):

protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21
form:
tablet
solubility:
water: soluble
storage temp.:
2-8°C

form

tablet

packaging

pkg of 30 tablets (individually packed in foil blister packs)

manufacturer/tradename

Roche

technique(s)

immunoblotting: suitable
western blot: suitable

solubility

water: soluble

absorption

0.4 at 280 nm (1x solution)

storage temp.

2-8°C

General description

Recombinant proteins may be partially degraded during expression. Soluble degradation products are co-purified and cause a non-homogeneous protein preparation with protein fragments of different lengths. However, metalloproteases are prevalent in (for example) bacteria and some techniques require that EDTA be omitted.

cOmplete ULTRA Tablets, Mini, EDTA-free contain the most powerful protease inhibitor cocktail for the optimal protection of your proteins against protease degradation. They are supplied as tablets in an EASYpack blister pack. EASYpacks provide the most convenient form of delivery - single tablets are easily pushed through the blister foil directly into the buffer. One cOmplete ULTRA Tablet, Mini, EDTA-free is sufficient for a volume of 10 ml extraction solution.

cOmplete ULTRA Tablets, Mini, EDTA-free are identical to cOmplete ULTRA Tablets, Mini, the only difference being that no EDTA or other chelating agents (e.g. EGTA) are included.

Specificity

Inhibits serine proteases, cysteine proteases, and aspartic proteases. Does not inhibit metalloproteases.

Application

During cell lysis and purification, target proteins are at risk of being cleaved by proteases, which are naturally present in cells. To ensure that full-length proteins are obtained, cOmplete ULTRA Tablets are used for the inhibition of an extended range of proteases. This new formulation shows highest inhibition ratios due to its broadened range of inhibitors compared to standard cOmplete Tablets.
This new protease inhibitor cocktail now inhibits aspartic proteases, as well as serine and cysteine proteases. cOmplete ULTRA Tablets were developed for maximum efficiency on different cell types. Best performance is guaranteed in lysates of bacteria, yeast, insect cells, and a wide range of higher cell lines.
This tablet does not contain EDTA; therefore, metalloproteases are not inhibited.
For the purification of histidine-tagged proteins using Immobilized Metal Ion Affinity Chromatography (IMAC), this EDTA-free tablet is recommended for applications using non-EDTA compatible IMAC resins. Alternatively, an EDTA-containing cOmplete ULTRA, Mini tablet may be used in combination with Roche ′s new EDTA-compatible chromatography material, cOmplete His-Tag Purification Resin.
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail has been used as a component of
  • homogenization buffer for muscle samples for immunoblotting sample preparation
  • radioimmunoprecipitation assay buffer (RIPA buffer) for protein extraction from brain samples for western blotting
  • lysis buffer for the human embryonic kidney 293 (HEK293) lysate preparation for western blotting

Features and Benefits

  • Protect your precious proteins.
Choose cOmplete ULTRA for even more efficient protease inhibition in eukaryotic, yeast, and bacterial cell lysates.
  • Why settle for less.
Benefit from 7 powerful protease inhibitors – more than in other EDTA-free inhibitor tablets.
  • Defend against a broad range of proteases.
Reliably inhibit serine, cysteine, and aspartic proteases.
  • Convenient and safe tablet format.
Eliminate weighing small amounts of individual inhibitors which might be hazardous. Simply drop a tablet from the blister into your buffers.

Components

Proprietary mixture of several protease inhibitors with broad inhibitory specificity.

Physical form

Tablets in foil blister packs. Each tablet is sufficient for a volume of 10 ml extraction solution.

Preparation Note

Working concentration: 1 tablet per 10 ml extraction solution
Working solution: Handling

Carefully push the tablet through the foil packaging using the base of your thumb (not your fingernail) to prevent the breakage of tablets.

Preparation of Working Solution

One tablet is sufficient for the inhibition of proteolytic activity in 10 ml extraction solution. If very high proteolytic activity is present, 2-3 tablets should be used for 10 ml extraction buffer. The tablets can be added directly to the extraction medium.

Stock Solution (2x conc.)

Alternatively to the direct use with lysates, a 2x stock solution can be prepared, with the following procedure:
• Add one cOmplete ULTRA Tablet, Mini, EDTA-free to 5 ml double-distilled water in a PE or PP tube.
• Vortex 3 min or until the tablet is completely dissolved.
• The stock solution is ready to use even if there are very fine parti­cles still visible in solution. These particles do not interfere with the inhibition performance of the tablets.
• This stock solution is stable for 4 weeks, stored at 2 to 8 °C, and for at least 4 weeks at -15 to -25 °C as well.
• During storage of the 2x stock solution, the particles can sediment. Briefly vortex the stock solution prior to use.

Storage conditions (working solution): The stock solution in water is stable for 4 weeks, stored at 2 to 8 °C, and for at least 4 weeks at  -15 to -25 °C as well.

Reconstitution

Soluble in aqueous buffers, or add directly to extraction media. For the stock solution, use water only.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

cOmplete is a trademark of Roche

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tony E R Werner et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(45), 27997-28004 (2020-10-24)
Amyloid formation involves the conversion of soluble protein species to an aggregated state. Amyloid fibrils of β-parvalbumin, a protein abundant in fish, act as an allergen but also inhibit the in vitro assembly of the Parkinson protein α-synuclein. However, the
Contrasting effects of individual versus combined estrogen and progestogen regimens as working memory load increases in middle-aged ovariectomized rats: one plus one does not equal two
Prakapenka AV, et al.
Neurobiology of Aging, 64, 1-14 (2018)
Blood flow restricted training leads to myocellular macrophage infiltration and upregulation of heat shock proteins, but no apparent muscle damage
Nielsen JL, et al.
The Journal of Physiology, 595(14), 4857-4873 (2017)
Bryan Frey et al.
Journal of neurochemistry, 157(4), 872-888 (2020-08-11)
Bimolecular fluorescence complementation (BiFC) was introduced a decade ago as a method to monitor alpha-synuclein (α-syn) oligomerization in intact cells. Since then, several α-syn BiFC cellular assays and animal models have been developed based on the assumption that an increase
Xiaofeng Guo et al.
Molecular cancer, 19(1), 83-83 (2020-05-08)
Accumulating evidence suggests that circular RNAs (circRNAs) are important participants in cancer progression. However, the biological processes and underlying mechanisms of circRNAs in pancreatic ductal adenocarcinoma (PDAC) are unclear. CircRNAs were verified by Sanger sequencing. Colony formation, 5-Ethynyl-2'-deoxyuridine (EdU), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service