Skip to Content
Merck
All Photos(2)

Key Documents

HPA039200

Sigma-Aldrich

Anti-ESYT3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Chr3syt, Anti-Extended synaptotagmin-like protein 3, Anti-Fam62c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PYADLTLEQRFQLDHSGLDSLISMRLVLRFLQVEERELGSPYTGPEALKKGPLLIKKVATNQGPKAQPQEEGPTDLPCPPDPASDTKD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ESYT3(83850)

General description

Extended synaptotagmin 3 (ESYT3) is encoded by the gene mapped to human chromosome 3q22.3. The encoded protein belongs to the family of evolutionarily conserved mammalian proteins, referred to as E-Syts. ESYT3 is localized to the plasma membrane (PM) and is characterized by an N-terminal transmembrane region, a conserved central juxtamembranous domain and three C-terminal C (2) domains.

Immunogen

extended synaptotagmin-like protein 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-ESYT3 antibody produced in rabbit has been used in immunoprecipitation assay.

Biochem/physiol Actions

Extended synaptotagmin 3 (ESYT3) plays a vital role under hypoxia conditions. Mutation in the gene contributes to the pathogenesis of coronary artery disease in humans.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79649

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PI(4,5)P(2)-dependent and Ca(2+)-regulated ER-PM interactions mediated by the extended synaptotagmins.
Giordano F
Cell, 153, 1494-1509 (2013)
Fine mapping of chromosome 3q22.3 identifies two haplotype blocks in ESYT3 associated with coronary artery disease in female Han Chinese.
Jiang F
Atherosclerosis, 218, 397-403 (2011)
E-Syts, a family of membranous Ca2+-sensor proteins with multiple C2 domains.
Min SW
Proceedings of the National Academy of Sciences of the USA, 104, 3823-3828 (2007)
Population Variation Revealed High-Altitude Adaptation of Tibetan Mastiffs
Li Y
Molecular Biology and Evolution, 31, 1200-1205 (2014)
Estela J Jauregui et al.
Biology of reproduction, 98(5), 722-738 (2018-02-07)
Spermatogenesis in mammals occurs in a very highly organized manner within the seminiferous epithelium regulated by different cell types in the testis. Testosterone produced by Leydig cells regulates blood-testis barrier formation, meiosis, spermiogenesis, and spermiation. However, it is unknown whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service