Skip to Content
Merck
All Photos(5)

Key Documents

HPA027878

Sigma-Aldrich

Anti-BAX antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BCL2-associated X protein, Anti-BCL2L4, Anti-GLUT10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GPTSSEQIMKTGALLLQGFIQDRAGRMGGEAPELALDPVPQDASTKKLSECLKRIGDELDSNMELQRMIAAVDTDSPREVFFRVAADMFSDGNFNW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

The BCL2 associated X, apoptosis regulator (BAX) gene has 6 exons and 5 intervening introns and is 6.939 bases long. Five protein-coding transcripts (α,ψ, β, δ, ε, σ) are found in the BAX gene. The BAX protein, a member of the B-cell lymphoma 2 (BCL2) family of proteins, can be found as a monomer, homodimer, or heterodimer. BAX is located in the mitochondria, mitochondrial outer membrane, endoplasmic reticulum membrane, mitochondrial permeability transition pore complex, and cytoplasm. BAX gene is located on human chromosome 19q13.33.

Immunogen

BCL2-associated X protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-BAX antibody produced in rabbit has been used in:
  • western blotting (1:100)
  • immunohistochemistry (1:50)
  • immunofluorescence (1:50)

Biochem/physiol Actions

The BCL2 associated X, apoptosis regulator (BAX) participates in the permeability pores formation that underlies necrosis. It heterodimerizes with the B-cell lymphoma 2 (BCL2) family members or tyrosine kinases to perform its proapoptotic role within the cell.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78050

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason Karch et al.
eLife, 2, e00772-e00772 (2013-08-31)
A critical event in ischemia-based cell death is the opening of the mitochondrial permeability transition pore (MPTP). However, the molecular identity of the components of the MPTP remains unknown. Here, we determined that the Bcl-2 family members Bax and Bak
Mona F Mahmoud et al.
Antioxidants (Basel, Switzerland), 11(3) (2022-03-26)
In the present study, we aimed to investigate the effect of coriander oil on dexamethasone-induced insulin resistance in rats and characterize its chemical composition using gas chromatography-mass spectrometry (GC-MS). Rats were divided into five groups (n = 6): Normal control
Tamas Vancsik et al.
Journal of Cancer, 9(1), 41-53 (2018-01-02)
Background: Modulated electro-hyperthermia (mEHT), a non-invasive intervention using 13.56 MHz radiofrequency, can selectively target cancers due to their elevated glycolysis (Warburg-effect), extracellular ion concentration and conductivity compared to normal tissues. We showed earlier that mEHT alone can provoke apoptosis and
Tetiana Kochubei et al.
Cell biology international, 40(12), 1313-1319 (2016-09-16)
Leukoagglutinin is one of the phytohemagglutinin isolectins isolated from Phaseolus vulgaris. In our recent study, we showed that this lectin is able to influence the growth of human cancer cells in vitro. In addition, using the acridine orange and ethidium
Colorectal cancer aggressiveness is related to fibronectin over expression, driving the activation of SDF-1: CXCR4 axis
Gouveia FS, et al.
International Journal of Cancer and Clinical Research, 3, 1-9 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service