Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501220

Sigma-Aldrich

Anti-GLP1R antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GLP-1 receptor, GLP-1-R, GLP-1R, GLP1R, Glucagon-like peptide 1 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 53 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLP1R(2740)

General description

Anti-GLP1R Antibody detects endogenous levels of total GLP1R protein.
Glucagon-like peptide 1 receptor (GLP1R) protein belongs to the class B G protein-coupled receptor (GPCR) secretin family. This classic seven-transmembrane protein is abundantly expressed in the cardiovascular system. The GLP1R gene is located on the human chromosome 6p21. Anti-GLP1R Antibody detects endogenous levels of total GLP1R protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human GLP1R.

Immunogen Range: 101-150

Application

Anti-GLP1R antibody produced in rabbit has been used in immunofluorescence staining.

Biochem/physiol Actions

Glucagon-like peptide 1 receptor (GLP1R) protein participates in the activation of protein kinase A (PKA), phosphoinositide 3-kinase (PI3K), MEK1/2, and endothelial nitric oxide synthase (eNOS) pathways. Hence, it plays a key role in apoptosis reduction, improved energy metabolism, decreased inflammation in myocardial cells, and vasodilation in myocardial arteries. The large N-terminal extracellular domain of GLP1R participates in ligand binding. Alterations in the GLP1R gene might increase the risk of coronary artery disease (CAD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sama Abdulmalik et al.
Differentiation; research in biological diversity, 120, 1-9 (2021-06-02)
Tendon injuries are common and account for up to 50% of musculoskeletal injuries in the United States. The poor healing nature of the tendon is attributed to poor vascularization and cellular composition. In the absence of FDA-approved growth factors for
Xiaowei Ma et al.
Journal of diabetes research, 2018, 1054192-1054192 (2018-10-03)
Glucagon-like peptide 1 (GLP-1) bestows protective effects upon the cardiovascular system through direct cardiovascular interactions or by improvements to metabolic function. Both these effects are thought to be at least partly mediated by the GLP-1 receptor (GLP-1R). This case-controlled study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service