Skip to Content
Merck
All Photos(6)

Key Documents

HPA024223

Sigma-Aldrich

Anti-MYO10 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Myosin-X, Anti-Unconventional myosin-10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

QRMKEQQELSLTEASLQKLQERRDQELRRLEEEACRAAQEFLESLNFDEIDECVRNIERSLSVGSEFSSELAESACEEKPNFNFSQPYPEEEVDEGFEADDDAFKDSPNPSEHGHSDQRTSGIRTSDDSSEEDPYMNDTVVPTSPSA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYO10(4651)

General description

The gene MYO10 (unconventional myosin-10) is mapped to human chromosome 5p15.1. It belongs to the MyTH4-FERM (myosin tail homology 4-band 4.1, ezrin, radixin, moesin, or MF) family of myosins. The protein has a myosin motor domain, a neck consisting of three IQ motifs and a tail region with three pleckstrin homology domains.

Immunogen

Myosin-X recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-MYO10 antibody produced in rabbit has ben used in western blotting and immunofluorescence.

Biochem/physiol Actions

MYO10 (unconventional myosin-10) is a microtubule-binding myosin. It is essential for centrosome positioning. It is a target gene for microRNA miR-340. In breast cancer, miR-340 inhibits invasion by silencing MYO10, which is responsible for aggressiveness and metastasis in breast cancer. The MYO10 gene is also a direct target of miR-124. Silencing of this gene suppresses cell invasion and metastasis in node-positive non-small cell lung cancer. MYO10 also plays a significant role in wound healing, invadopodia formation and angiogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85234

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NF-?B-mediated miR-124 suppresses metastasis of non-small-cell lung cancer by targeting MYO10.
Sun Y, et al.
Oncotarget, 6, 8244-8254 (2015)
Jeffrey Kroon et al.
Journal of immunology (Baltimore, Md. : 1950), 200(5), 1790-1801 (2018-02-02)
Leukocyte transendothelial migration is key to inflammation. Leukocytes first start rolling over the inflamed endothelium, followed by firmly adhering to it. Under inflammatory conditions, endothelial cells express small finger-like protrusions that stick out into the lumen. The function and regulation
John Caltagarone et al.
Molecular and cellular neurosciences, 68, 120-130 (2015-05-06)
Dopamine transporter (DAT, SLC6A3) controls dopamine (DA) neurotransmission by mediating re-uptake of extracellular DA into DA neurons. DA uptake depends on the amount of DAT at the cell surface, and is therefore regulated by DAT subcellular distribution. Hence we used
Direct Microtubule-Binding by Myosin-10 Orients Centrosomes toward Retraction Fibers and Subcortical Actin Clouds.
Kwon M, et al.
Developmental Cell, 34, 323-337 (2015)
Wan-Hsin Lin et al.
Journal of cell science, 126(Pt 20), 4756-4768 (2013-08-15)
Myosin X (Myo10) is an unconventional myosin with two known isoforms: full-length (FL)-Myo10 that has motor activity, and a recently identified brain-expressed isoform, headless (Hdl)-Myo10, which lacks most of the motor domain. FL-Myo10 is involved in the regulation of filopodia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service