Skip to Content
Merck
All Photos(3)

Key Documents

HPA018994

Sigma-Aldrich

Anti-KLK7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Kallikrein-7, Anti-Serine protease 6, Anti-Stratum corneum chymotryptic enzyme, Anti-hK7, Anti-hSCCE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 0.04-0.4 μg/mL

immunogen sequence

MVKKVRLPSRCEPPGTTCTVSGWGTTTSPDVTFPSDLMCVDVKLISPQDCTKVYKDLLENSMLCAGIPDSKKNACNGDSGGPLVCR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLK7(5650)

General description

The gene KLK7 (kallikrein-7) is mapped to human chromosome 19q13.4. It belongs to kallikrein-like gene family. KLKs are subgroup of serine proteases. KLK7 is expressed in skin, central nervous system, kidney, mammary and salivary glands. It is a secreted protein.

Immunogen

Kallikrein-7 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KLK7 (kallikrein-7) is responsible for cleavage of human insulin. Vaspin protein interacts with KLK7 and inhibits KLK7-mediated insulin degradation. Other substrates of KLK7 are midkine, CYR61 (cysteine-rich angiogenic inducer 61) and tenascin-C. It is also responsible for cleavage of procaspase-14 into an intermediate form, which is crucial for formation of mature form of caspase-14. KLK7 degrades intercellular cohesive structures in the outermost layer of the skin, thereby contributing to the cell shedding process at the skin surface.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74765

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John T Heiker et al.
Cellular and molecular life sciences : CMLS, 70(14), 2569-2583 (2013-02-02)
The molecular target of the adipokine vaspin (visceral adipose tissue-derived serpin; serpinA12) and its mode of action are unknown. Here, we provide the vaspin crystal structure and identify human kallikrein 7 (hK7) as a first protease target of vaspin inhibited
Mami Yamamoto et al.
The Journal of biological chemistry, 287(39), 32825-32834 (2012-07-25)
The maturation and activation mechanisms of caspases are generally well understood, except for those of caspase-14, which is activated at the onset of keratinocyte terminal differentiation. We investigated the possible involvement of epidermal proteases expressed in the late stage of
Ying Dong et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 9(5), 1710-1720 (2003-05-10)
The wild-type or variant mRNAs of several kallikrein (KLK) genes, such as KLK4, are highly expressed in ovarian carcinomas and may have potential as tumor markers. Two of these KLK genes (KLK5 and KLK7) and their proteins (hK5 and hK7)
G M Yousef et al.
Gene, 254(1-2), 119-128 (2000-09-07)
The human stratum corneum chymotryptic enzyme (HSCCE; PRSS6, KLK7 gene) has been purified from human stratum corneum and is known to participate in the cell shedding process. The cDNA of the gene has previously been reported. Here, we describe the
Yijing Yu et al.
The Journal of biological chemistry, 290(29), 17762-17775 (2015-06-03)
Kallikrein-related peptidases (KLKs) are a group of serine proteases widely expressed in various tissues and involved in a wide range of physiological and pathological processes. Although our understanding of the pathophysiological roles of most KLKs has blossomed in recent years

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service